Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
Chromosome Location
Chr.5: 140664799 - 140674119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC002482.2 - 7 - 8 1218 70

View Full Product Details