Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFWD3
Entrez Gene ID
55159
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
RNF201
Chromosome Location
Chr.16: 74621396 - 74666892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018124.3 NP_060594.3 3 - 4 825 77
XM_005256021.4 XP_005256078.1 4 - 5 958 77
XM_005256022.4 XP_005256079.1 3 - 4 1708 77
XM_006721228.3 XP_006721291.1 4 - 5 958 77
XM_011523191.2 XP_011521493.1 3 - 4 778 77
XM_017023391.1 XP_016878880.1 4 - 5 874 77
XM_017023392.1 XP_016878881.1 3 - 4 822 77
GenBank mRNA AK291200.1 - 4 - 5 925 77
BC002574.2 - 1 - 2 202 77
BC059371.1 - 3 - 4 774 77

View Full Product Details