The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003712.1 | NP_001003712.1 | 17 - 18 | 2350 | 108 |
NM_001319652.1 | NP_001306581.1 | 20 - 21 | 2475 | 108 | |
NM_001319653.1 | NP_001306582.1 | 18 - 19 | 2387 | 108 | |
NM_001319655.1 | NP_001306584.1 | 15 - 16 | 2281 | 108 | |
NM_020841.4 | NP_065892.1 | 18 - 19 | 2396 | 108 | |
XM_005268621.4 | XP_005268678.1 | 21 - 22 | 2494 | 108 | |
XM_006719224.2 | XP_006719287.1 | 17 - 18 | 2355 | 108 | |
XM_011537855.2 | XP_011536157.1 | 18 - 19 | 2013 | 108 | |
XM_011537856.2 | XP_011536158.1 | 19 - 20 | 2431 | 108 | |
XM_011537857.2 | XP_011536159.1 | 21 - 22 | 2485 | 108 | |
XM_017018768.1 | XP_016874257.1 | 18 - 19 | 2028 | 108 | |
XM_017018769.1 | XP_016874258.1 | 19 - 20 | 2399 | 108 | |
GenBank mRNA | AB040884.3 | - | 18 - 19 | 2067 | 108 |
AF392452.1 | - | 18 - 19 | 2397 | 108 | |
AK092160.1 | - | 15 - 16 | 2282 | 108 | |
AK289997.1 | - | 18 - 19 | 2116 | 108 | |
AL832876.1 | - | 5 - 6 | 452 | 108 | |
BC012935.1 | - | 4 - 5 | 381 | 108 | |
BC082235.1 | - | 18 - 19 | 2335 | 108 | |
BC093834.1 | - | 17 - 18 | 1995 | 108 | |
BC101529.1 | - | 17 - 18 | 1995 | 108 | |
BC111728.1 | - | 18 - 19 | 2290 | 108 | |
BX647211.1 | - | 17 - 18 | 1952 | 108 | |
CR749542.1 | - | 20 - 21 | 2200 | 108 |