Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAPLN2
Entrez Gene ID
60484
Gene Name
hyaluronan and proteoglycan link protein 2
Gene Aliases
BRAL1
Chromosome Location
Chr.1: 156601504 - 156626608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021817.2 NP_068589.1 5 - 6 958 70
XM_005245415.2 XP_005245472.1 5 - 6 927 70
XM_011509853.2 XP_011508155.1 5 - 6 1349 70
XM_017002020.1 XP_016857509.1 6 - 7 870 70
XM_017002021.1 XP_016857510.1 5 - 6 927 70
GenBank mRNA AB049054.1 - 5 - 6 958 70
AK090712.1 - 5 - 6 819 70
AK293797.1 - 4 - 5 928 70
AK294131.1 - 6 - 7 745 70
AK310758.1 - 3 - 4 1277 70
BC029864.1 - 5 - 6 755 70

View Full Product Details