Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RFWD2
- Entrez Gene ID
-
64326
- Gene Name
- ring finger and WD repeat domain 2
- Gene Aliases
-
COP1, RNF200
- Chromosome Location
-
Chr.1: 175944826 - 176207279 on Build GRCh38
View Full Product Details