The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000908.3 | NP_000899.1 | 8 - 8 | 5350 | 127 |
NM_001204375.1 | NP_001191304.1 | 8 - 8 | 5353 | 127 | |
NM_001204376.1 | NP_001191305.1 | 8 - 8 | 4283 | 127 | |
GenBank mRNA | AK024116.1 | - | 1 - 1 | 553 | 127 |
BC022250.1 | - | 1 - 1 | 789 | 127 |