The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322178.1 | NP_001309107.1 | 15 - 16 | 2833 | 144 |
NM_001322179.1 | NP_001309108.1 | 16 - 17 | 2983 | 144 | |
NM_001322180.1 | NP_001309109.1 | 15 - 16 | 2789 | 144 | |
NM_001322181.1 | NP_001309110.1 | 16 - 17 | 2980 | 144 | |
NM_001322187.1 | NP_001309116.1 | 17 - 18 | 3048 | 144 | |
NM_001322188.1 | NP_001309117.1 | 16 - 17 | 2983 | 144 | |
NM_001322189.1 | NP_001309118.1 | 17 - 18 | 3242 | 144 | |
NM_001322190.1 | NP_001309119.1 | 15 - 16 | 2839 | 144 | |
NM_001322191.1 | NP_001309120.1 | 15 - 16 | 2784 | 144 | |
NM_033401.4 | NP_207837.2 | 16 - 17 | 2983 | 144 | |
NM_138994.4 | NP_620481.2 | 15 - 16 | 2377 | 144 | |
NR_136210.1 | 16 - 17 | 2983 | 144 | ||
NR_136213.1 | 16 - 17 | 3024 | 144 | ||
NR_136216.1 | 18 - 19 | 3212 | 144 | ||
XM_011523403.2 | XP_011521705.1 | 16 - 17 | 2521 | 144 | |
XM_017023805.1 | XP_016879294.1 | 16 - 17 | 2712 | 144 | |
GenBank mRNA | AB051550.2 | - | 16 - 17 | 2922 | 144 |
AB100092.1 | - | 15 - 16 | 2393 | 144 | |
AB100093.1 | - | 14 - 15 | 2318 | 144 | |
AB100094.1 | - | 16 - 17 | 2537 | 144 | |
AF463518.1 | - | 15 - 16 | 2685 | 144 |