The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033550.3 | NP_291028.3 | 1 - 2 | 508 | 102 |
GenBank mRNA | AB017505.2 | - | 1 - 2 | 501 | 102 |
AB065434.1 | - | 1 - 2 | 530 | 102 | |
AK096502.1 | - | 1 - 2 | 289 | 102 | |
AK315182.1 | - | 1 - 2 | 334 | 102 | |
BC009727.1 | - | 1 - 2 | 438 | 102 | |
BC035461.1 | - | 1 - 2 | 282 | 102 | |
BC066309.1 | - | 1 - 2 | 304 | 102 | |
BM702732.1 | - | 1 - 2 | 509 | 103 |