Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 15200827-15201330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGMO
Entrez Gene ID
392636
Gene Name
alkylglycerol monooxygenase
Gene Aliases
TMEM195
UniGene
Hs.670634
Gene Chromosome Location
Chr.7: 15117223-15562015 on Build GRCh38
Transcripts
NM_001004320.1 XM_011515402.2 XM_017012204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6109508 c.1329T>A p.P443P Chr.7: 15201294-15201294 on Build GRCh38
3636566 c.1299T>C p.V433V Chr.7: 15201324-15201324 on Build GRCh38
351573 c.1295G>T p.G432V Chr.7: 15201328-15201328 on Build GRCh38
1087683 c.1304G>A p.S435N Chr.7: 15201319-15201319 on Build GRCh38
745811 c.1330T>A p.W444R Chr.7: 15201293-15201293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975972751 Chr.7: 15201167-15201167 on Build GRCh38
rs942093500 Chr.7: 15201227-15201227 on Build GRCh38
rs1040840636 Chr.7: 15200908-15200908 on Build GRCh38
rs878948423 Chr.7: 15201303-15201303 on Build GRCh38
rs141062961 Chr.7: 15201263-15201263 on Build GRCh38
rs909249683 Chr.7: 15200977-15200977 on Build GRCh38
rs548705604 Chr.7: 15201101-15201101 on Build GRCh38
rs901944024 Chr.7: 15200927-15200927 on Build GRCh38
rs752396046 Chr.7: 15201243-15201243 on Build GRCh38
rs142897311 Chr.7: 15200940-15200940 on Build GRCh38
rs1036326700 Chr.7: 15200984-15200984 on Build GRCh38
rs775943653 Chr.7: 15201255-15201256 on Build GRCh38
rs367795517 Chr.7: 15201240-15201240 on Build GRCh38
rs1030629781 Chr.7: 15201104-15201104 on Build GRCh38
rs946578341 Chr.7: 15201204-15201204 on Build GRCh38
rs935194123 Chr.7: 15200904-15200904 on Build GRCh38
rs745616103 Chr.7: 15201281-15201281 on Build GRCh38
rs746903538 Chr.7: 15201265-15201265 on Build GRCh38
rs953176363 Chr.7: 15201183-15201183 on Build GRCh38
rs529856304 Chr.7: 15201159-15201159 on Build GRCh38
rs151061395 Chr.7: 15201234-15201234 on Build GRCh38
rs375157677 Chr.7: 15201239-15201239 on Build GRCh38
rs960494026 Chr.7: 15201052-15201052 on Build GRCh38
rs764922677 Chr.7: 15201242-15201242 on Build GRCh38
rs930882645 Chr.7: 15200856-15200856 on Build GRCh38
rs1050505759 Chr.7: 15201179-15201179 on Build GRCh38
rs769593390 Chr.7: 15201295-15201295 on Build GRCh38
rs530852699 Chr.7: 15201034-15201034 on Build GRCh38
rs1011981115 Chr.7: 15201045-15201045 on Build GRCh38
rs544511923 Chr.7: 15201099-15201099 on Build GRCh38
rs562910582 Chr.7: 15200902-15200902 on Build GRCh38
rs907859648 Chr.7: 15200895-15200895 on Build GRCh38
rs1034692536 Chr.7: 15201257-15201257 on Build GRCh38
rs932072441 Chr.7: 15200951-15200951 on Build GRCh38
rs781143020 Chr.7: 15201280-15201280 on Build GRCh38
rs990945241 Chr.7: 15201066-15201066 on Build GRCh38
rs757838660 Chr.7: 15201248-15201248 on Build GRCh38
rs908141194 Chr.7: 15201118-15201118 on Build GRCh38
rs1049377599 Chr.7: 15200879-15200879 on Build GRCh38
rs894631040 Chr.7: 15201041-15201041 on Build GRCh38
rs1003053025 Chr.7: 15201001-15201001 on Build GRCh38
rs774371546 Chr.7: 15201297-15201297 on Build GRCh38
rs754491185 Chr.7: 15201195-15201195 on Build GRCh38
rs182789308 Chr.7: 15201070-15201070 on Build GRCh38
rs765529122 Chr.7: 15200871-15200871 on Build GRCh38
rs1013034945 Chr.7: 15201300-15201300 on Build GRCh38
rs923312103 Chr.7: 15201156-15201156 on Build GRCh38
rs186964232 Chr.7: 15201185-15201185 on Build GRCh38
rs1035151800 Chr.7: 15201015-15201015 on Build GRCh38
rs561105537 Chr.7: 15201133-15201133 on Build GRCh38
rs1050480979 Chr.7: 15200953-15200953 on Build GRCh38
rs35972659 Chr.7: 15200948-15200948 on Build GRCh38
rs757230298 Chr.7: 15201271-15201271 on Build GRCh38
rs567428292 Chr.7: 15201201-15201201 on Build GRCh38
rs909291674 Chr.7: 15201206-15201206 on Build GRCh38
rs916745315 Chr.7: 15201095-15201095 on Build GRCh38
rs534045528 Chr.7: 15201151-15201152 on Build GRCh38
rs940662883 Chr.7: 15201148-15201148 on Build GRCh38
rs565266503 Chr.7: 15200979-15200979 on Build GRCh38
rs765092483 Chr.7: 15201261-15201261 on Build GRCh38
rs898079828 Chr.7: 15200986-15200986 on Build GRCh38
rs1022199984 Chr.7: 15200959-15200959 on Build GRCh38
rs923850427 Chr.7: 15200899-15200899 on Build GRCh38
rs748470464 Chr.7: 15201302-15201302 on Build GRCh38
rs553197196 Chr.7: 15200916-15200916 on Build GRCh38
rs986273193 Chr.7: 15201202-15201202 on Build GRCh38
rs34708515 Chr.7: 15201153-15201154 on Build GRCh38
rs967777105 Chr.7: 15201096-15201096 on Build GRCh38
rs998394634 Chr.7: 15200928-15200928 on Build GRCh38
rs923122755 Chr.7: 15201175-15201175 on Build GRCh38

View Full Product Details