Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16600878-16601421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY2
Entrez Gene ID
57037
Gene Name
ankyrin repeat and MYND domain containing 2
Gene Aliases
ZMYND20
UniGene
Hs.157378
Gene Chromosome Location
Chr.7: 16599776-16645817 on Build GRCh38
Transcripts
NM_020319.2 XM_011515458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4460379 c.1163C>T p.S388F Chr.7: 16600924-16600924 on Build GRCh38
4461444 c.1206C>T p.I402I Chr.7: 16600881-16600881 on Build GRCh38
6109387 c.1187C>T p.P396L Chr.7: 16600900-16600900 on Build GRCh38
3306608 c.1171G>A p.V391I Chr.7: 16600916-16600916 on Build GRCh38
5716244 c.1207T>G p.S403A Chr.7: 16600880-16600880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549720703 Chr.7: 16601133-16601133 on Build GRCh38
rs775883371 Chr.7: 16600951-16600952 on Build GRCh38
rs930476181 Chr.7: 16601052-16601052 on Build GRCh38
rs781059731 Chr.7: 16601110-16601111 on Build GRCh38
rs1029511201 Chr.7: 16600935-16600935 on Build GRCh38
rs1028742835 Chr.7: 16601172-16601172 on Build GRCh38
rs201863855 Chr.7: 16600937-16600937 on Build GRCh38
rs543441975 Chr.7: 16601140-16601140 on Build GRCh38
rs374629071 Chr.7: 16600919-16600919 on Build GRCh38
rs866529971 Chr.7: 16601335-16601335 on Build GRCh38
rs780685529 Chr.7: 16600918-16600918 on Build GRCh38
rs781295552 Chr.7: 16601072-16601072 on Build GRCh38
rs906166634 Chr.7: 16601144-16601144 on Build GRCh38
rs571612432 Chr.7: 16601177-16601177 on Build GRCh38
rs541889372 Chr.7: 16601084-16601084 on Build GRCh38
rs567598349 Chr.7: 16600993-16600993 on Build GRCh38
rs960687950 Chr.7: 16601326-16601326 on Build GRCh38
rs1054470558 Chr.7: 16601167-16601167 on Build GRCh38
rs1006761508 Chr.7: 16601367-16601367 on Build GRCh38
rs745340130 Chr.7: 16601240-16601241 on Build GRCh38
rs371051652 Chr.7: 16601045-16601045 on Build GRCh38
rs891846289 Chr.7: 16601278-16601278 on Build GRCh38
rs780767662 Chr.7: 16601394-16601394 on Build GRCh38
rs746698447 Chr.7: 16600911-16600911 on Build GRCh38
rs77709309 Chr.7: 16601289-16601289 on Build GRCh38
rs966922877 Chr.7: 16601309-16601309 on Build GRCh38
rs935965025 Chr.7: 16601154-16601154 on Build GRCh38
rs758642076 Chr.7: 16600974-16600974 on Build GRCh38
rs750477693 Chr.7: 16601232-16601232 on Build GRCh38
rs770307031 Chr.7: 16600916-16600916 on Build GRCh38
rs990821590 Chr.7: 16601332-16601332 on Build GRCh38
rs796241399 Chr.7: 16600915-16600916 on Build GRCh38
rs748546862 Chr.7: 16601077-16601077 on Build GRCh38
rs957130309 Chr.7: 16601222-16601222 on Build GRCh38
rs1051639479 Chr.7: 16601356-16601356 on Build GRCh38
rs978708602 Chr.7: 16601311-16601311 on Build GRCh38
rs376628126 Chr.7: 16600965-16600965 on Build GRCh38
rs756634502 Chr.7: 16601336-16601336 on Build GRCh38
rs895270276 Chr.7: 16601031-16601031 on Build GRCh38
rs533394063 Chr.7: 16600957-16600959 on Build GRCh38
rs760121038 Chr.7: 16600955-16600955 on Build GRCh38
rs773263711 Chr.7: 16600933-16600933 on Build GRCh38
rs770252218 Chr.7: 16601321-16601321 on Build GRCh38
rs1022856069 Chr.7: 16601256-16601256 on Build GRCh38
rs1055100507 Chr.7: 16600996-16600996 on Build GRCh38
rs551624387 Chr.7: 16600907-16600907 on Build GRCh38
rs1011063340 Chr.7: 16601254-16601254 on Build GRCh38
rs764553055 Chr.7: 16600983-16600983 on Build GRCh38
rs553963594 Chr.7: 16601237-16601237 on Build GRCh38
rs890949364 Chr.7: 16601361-16601361 on Build GRCh38
rs1005181346 Chr.7: 16601076-16601076 on Build GRCh38
rs886347482 Chr.7: 16601074-16601074 on Build GRCh38
rs983995769 Chr.7: 16601109-16601109 on Build GRCh38
rs753092098 Chr.7: 16600973-16600973 on Build GRCh38
rs928245130 Chr.7: 16601325-16601325 on Build GRCh38
rs368634987 Chr.7: 16600927-16600927 on Build GRCh38
rs917441419 Chr.7: 16601131-16601131 on Build GRCh38
rs538921061 Chr.7: 16601207-16601207 on Build GRCh38
rs140918019 Chr.7: 16600923-16600923 on Build GRCh38
rs200413443 Chr.7: 16600943-16600943 on Build GRCh38
rs973774069 Chr.7: 16601019-16601019 on Build GRCh38
rs202049267 Chr.7: 16600944-16600944 on Build GRCh38
rs145736260 Chr.7: 16601314-16601314 on Build GRCh38

View Full Product Details