Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ODF3
Entrez Gene ID
113746
Gene Name
outer dense fiber of sperm tails 3
Gene Aliases
CT135, SHIPPO1, TISP50
Chromosome Location
Chr.11: 196761 - 200258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286136.1 NP_001273065.1 1 - 2 284 60
NM_053280.4 NP_444510.2 1 - 2 284 60
GenBank mRNA AB067774.1 - 1 - 2 391 60
AY713299.1 - 1 - 2 259 60
BC126222.1 - 1 - 2 92 60
BC126248.1 - 1 - 2 92 60
BC144022.1 - 1 - 2 92 60

View Full Product Details