The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080476.4 | NP_536724.1 | 5 - 6 | 624 | 88 |
XM_017027664.1 | XP_016883153.1 | 5 - 6 | 442 | 88 | |
XM_017027665.1 | XP_016883154.1 | 5 - 6 | 442 | 88 | |
GenBank mRNA | AB086842.1 | - | 5 - 6 | 430 | 88 |
AK075507.1 | - | 5 - 6 | 449 | 88 | |
AK301900.1 | - | 5 - 6 | 424 | 88 | |
AK308627.1 | - | 4 - 5 | 371 | 88 | |
AY339061.1 | - | 4 - 5 | 364 | 88 | |
AY358816.1 | - | 5 - 6 | 454 | 88 | |
AY422169.1 | - | 5 - 6 | 624 | 88 | |
BC030512.1 | - | 5 - 6 | 435 | 88 |