Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81702026-81702560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

SNP Detail

SNP ID Chromosome Location
rs992946888 Chr.7: 81702392-81702392 on Build GRCh38
rs899567940 Chr.7: 81702218-81702218 on Build GRCh38
rs182057547 Chr.7: 81702185-81702185 on Build GRCh38
rs535633453 Chr.7: 81702167-81702167 on Build GRCh38
rs926214806 Chr.7: 81702134-81702134 on Build GRCh38
rs374789745 Chr.7: 81702478-81702479 on Build GRCh38
rs573406409 Chr.7: 81702515-81702515 on Build GRCh38
rs890647502 Chr.7: 81702175-81702175 on Build GRCh38
rs1053369698 Chr.7: 81702331-81702331 on Build GRCh38
rs886062456 Chr.7: 81702480-81702480 on Build GRCh38
rs750490813 Chr.7: 81702533-81702533 on Build GRCh38
rs934801656 Chr.7: 81702148-81702148 on Build GRCh38
rs531222230 Chr.7: 81702413-81702413 on Build GRCh38
rs879724696 Chr.7: 81702238-81702238 on Build GRCh38
rs1051166277 Chr.7: 81702243-81702243 on Build GRCh38
rs977727635 Chr.7: 81702493-81702493 on Build GRCh38
rs539702697 Chr.7: 81702357-81702357 on Build GRCh38
rs994551612 Chr.7: 81702204-81702204 on Build GRCh38
rs537468151 Chr.7: 81702228-81702228 on Build GRCh38
rs1038792244 Chr.7: 81702187-81702187 on Build GRCh38
rs934877141 Chr.7: 81702330-81702330 on Build GRCh38
rs5745768 Chr.7: 81702114-81702114 on Build GRCh38
rs545984462 Chr.7: 81702264-81702267 on Build GRCh38
rs945946659 Chr.7: 81702111-81702111 on Build GRCh38
rs777313117 Chr.7: 81702322-81702322 on Build GRCh38
rs1034775459 Chr.7: 81702272-81702273 on Build GRCh38
rs878879830 Chr.7: 81702276-81702276 on Build GRCh38
rs960578259 Chr.7: 81702273-81702273 on Build GRCh38
rs568550366 Chr.7: 81702194-81702194 on Build GRCh38
rs758013381 Chr.7: 81702309-81702309 on Build GRCh38
rs1004774004 Chr.7: 81702261-81702261 on Build GRCh38
rs566500466 Chr.7: 81702118-81702118 on Build GRCh38
rs577625800 Chr.7: 81702315-81702315 on Build GRCh38
rs542402530 Chr.7: 81702524-81702524 on Build GRCh38
rs958744890 Chr.7: 81702379-81702379 on Build GRCh38
rs930299625 Chr.7: 81702399-81702399 on Build GRCh38
rs5745769 Chr.7: 81702057-81702058 on Build GRCh38
rs778665027 Chr.7: 81702129-81702129 on Build GRCh38
rs1027510708 Chr.7: 81702213-81702213 on Build GRCh38
rs993314088 Chr.7: 81702274-81702274 on Build GRCh38
rs754673292 Chr.7: 81702059-81702059 on Build GRCh38
rs567921457 Chr.7: 81702054-81702055 on Build GRCh38
rs935865465 Chr.7: 81702135-81702135 on Build GRCh38
rs763357973 Chr.7: 81702314-81702315 on Build GRCh38
rs886820277 Chr.7: 81702263-81702263 on Build GRCh38
rs900161616 Chr.7: 81702198-81702198 on Build GRCh38
rs186434043 Chr.7: 81702279-81702279 on Build GRCh38
rs765177187 Chr.7: 81702532-81702532 on Build GRCh38
rs767556298 Chr.7: 81702536-81702536 on Build GRCh38
rs933743740 Chr.7: 81702219-81702219 on Build GRCh38
rs533030741 Chr.7: 81702089-81702089 on Build GRCh38
rs917265278 Chr.7: 81702067-81702067 on Build GRCh38
rs544879896 Chr.7: 81702360-81702360 on Build GRCh38
rs1024724608 Chr.7: 81702422-81702422 on Build GRCh38
rs970443569 Chr.7: 81702313-81702313 on Build GRCh38
rs912015642 Chr.7: 81702351-81702351 on Build GRCh38
rs967428555 Chr.7: 81702462-81702462 on Build GRCh38

View Full Product Details