The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130901.1 | NP_570971.1 | 7 - 8 | 1095 | 83 |
GenBank mRNA | AJ430383.1 | - | 7 - 8 | 1095 | 83 |
BC035668.1 | - | 9 - 10 | 1192 | 83 |