Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPACA4
Entrez Gene ID
171169
Gene Name
sperm acrosome associated 4
Gene Aliases
SAMP14
Chromosome Location
Chr.19: 48606743 - 48607714 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133498.2 NP_598005.1 1 - 1 100 75
GenBank mRNA AF353721.2 - 1 - 1 100 75
AY358387.1 - 1 - 1 82 75
BC047334.1 - 1 - 1 80 75

View Full Product Details