Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYGB
Entrez Gene ID
114757
Gene Name
cytoglobin
Gene Aliases
HGB, STAP
Chromosome Location
Chr.17: 76527348 - 76557692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134268.4 NP_599030.1 1 - 2 509 97
XM_005257005.2 XP_005257062.1 1 - 2 156 97
GenBank mRNA AB057769.1 - 1 - 2 189 97
AJ315162.1 - 1 - 2 146 97
AK098057.1 - 1 - 2 156 97
BQ881744.1 - 1 - 2 509 97

View Full Product Details