The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138295.3 | NP_612152.1 | 28 - 29 | 4628 | 70 |
XM_017011798.1 | XP_016867287.1 | 29 - 30 | 11033 | 70 | |
GenBank mRNA | AB061683.1 | - | 28 - 29 | 4594 | 70 |