The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138392.3 | NP_612401.2 | 17 - 18 | 1943 | 86 |
XM_006723474.1 | XP_006723537.1 | 17 - 18 | 1973 | 86 | |
XM_006723475.1 | XP_006723538.1 | 17 - 18 | 1970 | 86 | |
XM_011527492.1 | XP_011525794.1 | 14 - 15 | 1622 | 86 | |
XM_011527493.1 | XP_011525795.1 | 16 - 17 | 1869 | 86 | |
XM_017027473.1 | XP_016882962.1 | 17 - 18 | 1940 | 86 | |
XM_017027474.1 | XP_016882963.1 | 16 - 17 | 1839 | 86 | |
GenBank mRNA | AF258553.1 | - | 17 - 18 | 1942 | 86 |
AK075057.1 | - | 17 - 18 | 1865 | 86 | |
AK123695.1 | - | 15 - 16 | 1921 | 86 | |
AK297006.1 | - | 11 - 12 | 1569 | 86 | |
AK300819.1 | - | 16 - 17 | 1722 | 86 | |
AK312748.1 | - | 17 - 18 | 1907 | 86 | |
AY007117.1 | - | 8 - 9 | 978 | 86 | |
BC007653.1 | - | 9 - 10 | 1229 | 86 | |
BC022855.1 | - | 17 - 18 | 1914 | 86 | |
CA416700.1 | - | 1 - 2 | 86 |