Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91991817-91992320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP9
Entrez Gene ID
10142
Gene Name
A-kinase anchoring protein 9
Gene Aliases
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
UniGene
Hs.651221
Gene Chromosome Location
Chr.7: 91940867-92110673 on Build GRCh38
Transcripts
NM_005751.4 NM_147185.2 XM_017011642.1 XM_017011643.1 XM_017011644.1 XM_017011645.1 XM_017011646.1 XM_017011647.1 XM_017011648.1 XM_017011649.1 XM_017011650.1 XM_017011651.1 XM_017011652.1 XM_017011653.1 XM_017011654.1 XM_017011655.1 XM_017011656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2863564 c.397T>G p.L133V Chr.7: 91992203-91992203 on Build GRCh38
1550358 c.405G>T p.R135S Chr.7: 91992211-91992211 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775043801 Chr.7: 91992244-91992244 on Build GRCh38
rs777124971 Chr.7: 91992164-91992164 on Build GRCh38
rs576155671 Chr.7: 91991870-91991870 on Build GRCh38
rs1008827202 Chr.7: 91991943-91991943 on Build GRCh38
rs565231706 Chr.7: 91992075-91992075 on Build GRCh38
rs757027446 Chr.7: 91992212-91992212 on Build GRCh38
rs886471976 Chr.7: 91992229-91992229 on Build GRCh38
rs541958837 Chr.7: 91991878-91991878 on Build GRCh38
rs186249152 Chr.7: 91992045-91992045 on Build GRCh38
rs773227815 Chr.7: 91992159-91992159 on Build GRCh38
rs755049262 Chr.7: 91992200-91992200 on Build GRCh38
rs1044552002 Chr.7: 91991879-91991879 on Build GRCh38
rs745476087 Chr.7: 91992113-91992113 on Build GRCh38
rs567864641 Chr.7: 91992142-91992142 on Build GRCh38
rs764576466 Chr.7: 91992162-91992162 on Build GRCh38
rs964870415 Chr.7: 91991919-91991919 on Build GRCh38
rs770693862 Chr.7: 91992107-91992107 on Build GRCh38
rs774072518 Chr.7: 91992043-91992043 on Build GRCh38
rs993058903 Chr.7: 91992083-91992083 on Build GRCh38
rs375975524 Chr.7: 91992125-91992125 on Build GRCh38
rs746539511 Chr.7: 91992249-91992249 on Build GRCh38
rs954922486 Chr.7: 91992002-91992002 on Build GRCh38
rs541483705 Chr.7: 91991909-91991909 on Build GRCh38
rs917845186 Chr.7: 91992124-91992124 on Build GRCh38
rs761240315 Chr.7: 91992146-91992146 on Build GRCh38
rs772352621 Chr.7: 91991948-91991948 on Build GRCh38
rs771710726 Chr.7: 91992243-91992243 on Build GRCh38
rs527776244 Chr.7: 91991904-91991904 on Build GRCh38
rs758661269 Chr.7: 91992194-91992194 on Build GRCh38
rs925001863 Chr.7: 91992060-91992060 on Build GRCh38
rs1048100179 Chr.7: 91992133-91992133 on Build GRCh38
rs751675949 Chr.7: 91992198-91992198 on Build GRCh38
rs778747883 Chr.7: 91992216-91992216 on Build GRCh38
rs754335465 Chr.7: 91992178-91992178 on Build GRCh38
rs549302363 Chr.7: 91992091-91992091 on Build GRCh38
rs1014013106 Chr.7: 91991856-91991856 on Build GRCh38
rs562168122 Chr.7: 91991893-91991893 on Build GRCh38
rs773907433 Chr.7: 91992110-91992110 on Build GRCh38
rs746659583 Chr.7: 91992030-91992030 on Build GRCh38
rs200518712 Chr.7: 91992195-91992195 on Build GRCh38
rs779551997 Chr.7: 91991902-91991902 on Build GRCh38
rs116169457 Chr.7: 91991952-91991952 on Build GRCh38
rs4364573 Chr.7: 91992003-91992003 on Build GRCh38
rs749209057 Chr.7: 91992211-91992211 on Build GRCh38
rs750656682 Chr.7: 91992186-91992186 on Build GRCh38
rs1026277942 Chr.7: 91991889-91991889 on Build GRCh38
rs376736925 Chr.7: 91992256-91992256 on Build GRCh38
rs188318223 Chr.7: 91992179-91992179 on Build GRCh38
rs1006342445 Chr.7: 91991906-91991906 on Build GRCh38
rs371461215 Chr.7: 91992294-91992294 on Build GRCh38
rs528086679 Chr.7: 91992035-91992035 on Build GRCh38
rs550423073 Chr.7: 91992021-91992021 on Build GRCh38
rs979232473 Chr.7: 91992056-91992056 on Build GRCh38
rs1008881176 Chr.7: 91992275-91992275 on Build GRCh38
rs974856812 Chr.7: 91991924-91991924 on Build GRCh38
rs776175151 Chr.7: 91992074-91992074 on Build GRCh38
rs142247871 Chr.7: 91991842-91991842 on Build GRCh38
rs765586191 Chr.7: 91992184-91992184 on Build GRCh38
rs1022118256 Chr.7: 91992024-91992024 on Build GRCh38
rs964595638 Chr.7: 91991985-91991985 on Build GRCh38

View Full Product Details