Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 98291019-98291532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRI3
Entrez Gene ID
25798
Gene Name
brain protein I3
Gene Aliases
I3
UniGene
Hs.567438
Gene Chromosome Location
Chr.7: 98281667-98322398 on Build GRCh38
Transcripts
NM_001159491.1 NM_015379.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2866301 c.328T>G p.F110V Chr.7: 98291193-98291193 on Build GRCh38
7283979 c.346C>T p.R116* Chr.7: 98291211-98291211 on Build GRCh38
3833566 c.347G>A p.R116Q Chr.7: 98291212-98291212 on Build GRCh38
1568789 c.369C>T p.T123T Chr.7: 98291234-98291234 on Build GRCh38
1644437 c.285C>G p.I95M Chr.7: 98291150-98291150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014472674 Chr.7: 98291367-98291367 on Build GRCh38
rs765776994 Chr.7: 98291138-98291149 on Build GRCh38
rs370157248 Chr.7: 98291078-98291078 on Build GRCh38
rs770923573 Chr.7: 98291068-98291068 on Build GRCh38
rs761977710 Chr.7: 98291132-98291132 on Build GRCh38
rs781673839 Chr.7: 98291165-98291165 on Build GRCh38
rs925936861 Chr.7: 98291466-98291466 on Build GRCh38
rs772371426 Chr.7: 98291282-98291283 on Build GRCh38
rs1046980762 Chr.7: 98291372-98291372 on Build GRCh38
rs201437729 Chr.7: 98291069-98291069 on Build GRCh38
rs951039166 Chr.7: 98291178-98291178 on Build GRCh38
rs1016603202 Chr.7: 98291390-98291390 on Build GRCh38
rs774581185 Chr.7: 98291213-98291213 on Build GRCh38
rs925902801 Chr.7: 98291101-98291101 on Build GRCh38
rs1052229861 Chr.7: 98291045-98291045 on Build GRCh38
rs937413577 Chr.7: 98291347-98291347 on Build GRCh38
rs1045473019 Chr.7: 98291144-98291144 on Build GRCh38
rs375398915 Chr.7: 98291260-98291260 on Build GRCh38
rs895666656 Chr.7: 98291360-98291360 on Build GRCh38
rs1029775844 Chr.7: 98291302-98291302 on Build GRCh38
rs913213770 Chr.7: 98291055-98291055 on Build GRCh38
rs746969865 Chr.7: 98291254-98291254 on Build GRCh38
rs11539230 Chr.7: 98291154-98291154 on Build GRCh38
rs770581415 Chr.7: 98291184-98291184 on Build GRCh38
rs766891910 Chr.7: 98291086-98291095 on Build GRCh38
rs771124369 Chr.7: 98291271-98291271 on Build GRCh38
rs367759325 Chr.7: 98291281-98291281 on Build GRCh38
rs760184789 Chr.7: 98291089-98291098 on Build GRCh38
rs774390971 Chr.7: 98291120-98291120 on Build GRCh38
rs903152431 Chr.7: 98291465-98291465 on Build GRCh38
rs529949746 Chr.7: 98291393-98291393 on Build GRCh38
rs954914784 Chr.7: 98291406-98291406 on Build GRCh38
rs751815687 Chr.7: 98291281-98291282 on Build GRCh38
rs755471520 Chr.7: 98291088-98291088 on Build GRCh38
rs7015 Chr.7: 98291311-98291311 on Build GRCh38
rs11539224 Chr.7: 98291224-98291224 on Build GRCh38
rs112008968 Chr.7: 98291098-98291098 on Build GRCh38
rs140237370 Chr.7: 98291123-98291123 on Build GRCh38
rs535071936 Chr.7: 98291159-98291159 on Build GRCh38
rs926072460 Chr.7: 98291345-98291345 on Build GRCh38
rs564856231 Chr.7: 98291349-98291349 on Build GRCh38
rs949700849 Chr.7: 98291127-98291127 on Build GRCh38
rs765487035 Chr.7: 98291079-98291079 on Build GRCh38
rs1029802898 Chr.7: 98291398-98291398 on Build GRCh38
rs773385746 Chr.7: 98291211-98291211 on Build GRCh38
rs376999715 Chr.7: 98291404-98291404 on Build GRCh38
rs548377152 Chr.7: 98291458-98291458 on Build GRCh38
rs571655703 Chr.7: 98291418-98291418 on Build GRCh38
rs998227361 Chr.7: 98291422-98291422 on Build GRCh38
rs957243173 Chr.7: 98291474-98291474 on Build GRCh38
rs780177079 Chr.7: 98291253-98291253 on Build GRCh38
rs771966921 Chr.7: 98291206-98291206 on Build GRCh38
rs772940616 Chr.7: 98291063-98291064 on Build GRCh38
rs1056057547 Chr.7: 98291348-98291348 on Build GRCh38
rs772300526 Chr.7: 98291212-98291212 on Build GRCh38
rs9190 Chr.7: 98291251-98291251 on Build GRCh38
rs142425398 Chr.7: 98291237-98291237 on Build GRCh38
rs750648700 Chr.7: 98291138-98291138 on Build GRCh38
rs368070657 Chr.7: 98291088-98291089 on Build GRCh38
rs886983901 Chr.7: 98291379-98291379 on Build GRCh38
rs754616183 Chr.7: 98291495-98291495 on Build GRCh38
rs776866707 Chr.7: 98291131-98291131 on Build GRCh38
rs200497431 Chr.7: 98291259-98291259 on Build GRCh38
rs1033489475 Chr.7: 98291470-98291470 on Build GRCh38
rs915802849 Chr.7: 98291371-98291371 on Build GRCh38
rs755453031 Chr.7: 98291163-98291163 on Build GRCh38
rs750881831 Chr.7: 98291082-98291082 on Build GRCh38
rs780656170 Chr.7: 98291085-98291085 on Build GRCh38
rs991599992 Chr.7: 98291502-98291502 on Build GRCh38
rs753086498 Chr.7: 98291146-98291147 on Build GRCh38
rs760861800 Chr.7: 98291071-98291071 on Build GRCh38
rs867982825 Chr.7: 98291197-98291197 on Build GRCh38
rs553049612 Chr.7: 98291198-98291198 on Build GRCh38
rs765610787 Chr.7: 98291137-98291137 on Build GRCh38
rs759868427 Chr.7: 98291283-98291283 on Build GRCh38
rs748912411 Chr.7: 98291171-98291171 on Build GRCh38
rs191894782 Chr.7: 98291363-98291363 on Build GRCh38
rs998953618 Chr.7: 98291468-98291470 on Build GRCh38
rs773189720 Chr.7: 98291366-98291367 on Build GRCh38
rs778598033 Chr.7: 98291186-98291186 on Build GRCh38
rs145197209 Chr.7: 98291207-98291207 on Build GRCh38
rs190137205 Chr.7: 98291048-98291048 on Build GRCh38
rs559898953 Chr.7: 98291260-98291260 on Build GRCh38
rs762670783 Chr.7: 98291065-98291065 on Build GRCh38
rs752273619 Chr.7: 98291166-98291166 on Build GRCh38
rs111602129 Chr.7: 98291331-98291331 on Build GRCh38
rs62479857 Chr.7: 98291388-98291388 on Build GRCh38
rs9327 Chr.7: 98291289-98291289 on Build GRCh38
rs777670963 Chr.7: 98291277-98291278 on Build GRCh38
rs1020532050 Chr.7: 98291419-98291419 on Build GRCh38
rs371709705 Chr.7: 98291290-98291290 on Build GRCh38
rs930166898 Chr.7: 98291225-98291225 on Build GRCh38
rs941501643 Chr.7: 98291416-98291416 on Build GRCh38
rs977693263 Chr.7: 98291315-98291315 on Build GRCh38
rs183639368 Chr.7: 98291384-98291384 on Build GRCh38
rs143355417 Chr.7: 98291238-98291238 on Build GRCh38
rs751978646 Chr.7: 98291086-98291086 on Build GRCh38
rs758186767 Chr.7: 98291249-98291249 on Build GRCh38
rs764052604 Chr.7: 98291073-98291073 on Build GRCh38
rs376298920 Chr.7: 98291074-98291074 on Build GRCh38
rs766685450 Chr.7: 98291150-98291150 on Build GRCh38
rs188979881 Chr.7: 98291387-98291387 on Build GRCh38
rs770609756 Chr.7: 98291114-98291114 on Build GRCh38
rs991720823 Chr.7: 98291488-98291488 on Build GRCh38
rs569949932 Chr.7: 98291084-98291084 on Build GRCh38
rs373993202 Chr.7: 98291097-98291097 on Build GRCh38
rs775927267 Chr.7: 98291308-98291309 on Build GRCh38
rs749140392 Chr.7: 98291105-98291105 on Build GRCh38
rs1043034848 Chr.7: 98291340-98291340 on Build GRCh38
rs749321421 Chr.7: 98291268-98291268 on Build GRCh38
rs371589940 Chr.7: 98291061-98291061 on Build GRCh38
rs747598019 Chr.7: 98291095-98291095 on Build GRCh38
rs780588396 Chr.7: 98291094-98291094 on Build GRCh38
rs967594943 Chr.7: 98291456-98291456 on Build GRCh38
rs998157328 Chr.7: 98291291-98291291 on Build GRCh38
rs531028404 Chr.7: 98291146-98291146 on Build GRCh38
rs760364578 Chr.7: 98291083-98291085 on Build GRCh38
rs563450266 Chr.7: 98291464-98291464 on Build GRCh38
rs748283835 Chr.7: 98291208-98291208 on Build GRCh38
rs759942845 Chr.7: 98291227-98291227 on Build GRCh38
rs777451732 Chr.7: 98291168-98291168 on Build GRCh38
rs778563786 Chr.7: 98291244-98291244 on Build GRCh38
rs957395659 Chr.7: 98291337-98291337 on Build GRCh38
rs767968624 Chr.7: 98291235-98291235 on Build GRCh38
rs996593100 Chr.7: 98291394-98291394 on Build GRCh38
rs12865 Chr.7: 98291232-98291232 on Build GRCh38
rs137943501 Chr.7: 98291209-98291209 on Build GRCh38
rs557262739 Chr.7: 98291245-98291245 on Build GRCh38
rs145484995 Chr.7: 98291121-98291121 on Build GRCh38
rs771070435 Chr.7: 98291423-98291423 on Build GRCh38

View Full Product Details