Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 98306206-98306701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2L1
Entrez Gene ID
55971
Gene Name
BAI1 associated protein 2 like 1
Gene Aliases
IRTKS
UniGene
Hs.656063
Gene Chromosome Location
Chr.7: 98291650-98401115 on Build GRCh38
Transcripts
NM_018842.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Gene Symbol
BRI3
Entrez Gene ID
25798
Gene Name
brain protein I3
Gene Aliases
I3
UniGene
Hs.567438
Gene Chromosome Location
Chr.7: 98281667-98322398 on Build GRCh38
Transcripts
NM_018842.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193529 c.1227C>T p.T409T Chr.7: 98306453-98306453 on Build GRCh38
1093518 c.1191G>A p.T397T Chr.7: 98306489-98306489 on Build GRCh38
6875472 c.1241G>A p.S414N Chr.7: 98306439-98306439 on Build GRCh38
5475104 c.1178C>T p.P393L Chr.7: 98306502-98306502 on Build GRCh38
238938 c.1173G>C p.W391C Chr.7: 98306507-98306507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008716029 Chr.7: 98306307-98306307 on Build GRCh38
rs200799373 Chr.7: 98306566-98306566 on Build GRCh38
rs371518872 Chr.7: 98306411-98306411 on Build GRCh38
rs1041664222 Chr.7: 98306354-98306354 on Build GRCh38
rs765507923 Chr.7: 98306363-98306363 on Build GRCh38
rs1000079497 Chr.7: 98306360-98306360 on Build GRCh38
rs762651885 Chr.7: 98306511-98306511 on Build GRCh38
rs13232181 Chr.7: 98306359-98306359 on Build GRCh38
rs775956056 Chr.7: 98306398-98306398 on Build GRCh38
rs769259940 Chr.7: 98306590-98306590 on Build GRCh38
rs1008074554 Chr.7: 98306614-98306614 on Build GRCh38
rs748157748 Chr.7: 98306558-98306559 on Build GRCh38
rs760146100 Chr.7: 98306405-98306405 on Build GRCh38
rs542738993 Chr.7: 98306282-98306282 on Build GRCh38
rs763694432 Chr.7: 98306410-98306410 on Build GRCh38
rs111919043 Chr.7: 98306301-98306301 on Build GRCh38
rs958358104 Chr.7: 98306379-98306379 on Build GRCh38
rs776978477 Chr.7: 98306595-98306595 on Build GRCh38
rs553626222 Chr.7: 98306626-98306626 on Build GRCh38
rs758242917 Chr.7: 98306430-98306430 on Build GRCh38
rs182223141 Chr.7: 98306600-98306600 on Build GRCh38
rs1011479666 Chr.7: 98306640-98306640 on Build GRCh38
rs771725767 Chr.7: 98306651-98306651 on Build GRCh38
rs553839402 Chr.7: 98306260-98306260 on Build GRCh38
rs565003158 Chr.7: 98306499-98306499 on Build GRCh38
rs1038359368 Chr.7: 98306568-98306568 on Build GRCh38
rs982702472 Chr.7: 98306447-98306447 on Build GRCh38
rs142825458 Chr.7: 98306281-98306281 on Build GRCh38
rs138564547 Chr.7: 98306498-98306498 on Build GRCh38
rs775794764 Chr.7: 98306486-98306486 on Build GRCh38
rs754822606 Chr.7: 98306450-98306450 on Build GRCh38
rs768288557 Chr.7: 98306387-98306387 on Build GRCh38
rs754008612 Chr.7: 98306547-98306547 on Build GRCh38
rs779147963 Chr.7: 98306559-98306559 on Build GRCh38
rs554579335 Chr.7: 98306353-98306353 on Build GRCh38
rs764973996 Chr.7: 98306426-98306426 on Build GRCh38
rs989829844 Chr.7: 98306442-98306442 on Build GRCh38
rs955959958 Chr.7: 98306635-98306635 on Build GRCh38
rs746122454 Chr.7: 98306565-98306565 on Build GRCh38
rs1038123758 Chr.7: 98306264-98306264 on Build GRCh38
rs780179039 Chr.7: 98306444-98306444 on Build GRCh38
rs746282005 Chr.7: 98306471-98306471 on Build GRCh38
rs750590387 Chr.7: 98306395-98306396 on Build GRCh38
rs1004171208 Chr.7: 98306302-98306302 on Build GRCh38
rs761545153 Chr.7: 98306415-98306415 on Build GRCh38
rs890332505 Chr.7: 98306306-98306306 on Build GRCh38
rs866394377 Chr.7: 98306476-98306476 on Build GRCh38
rs973646026 Chr.7: 98306502-98306502 on Build GRCh38
rs1012660837 Chr.7: 98306382-98306382 on Build GRCh38
rs754827998 Chr.7: 98306403-98306403 on Build GRCh38
rs777922061 Chr.7: 98306452-98306452 on Build GRCh38
rs112997351 Chr.7: 98306375-98306375 on Build GRCh38
rs997374248 Chr.7: 98306581-98306581 on Build GRCh38
rs766297356 Chr.7: 98306521-98306521 on Build GRCh38
rs1021327861 Chr.7: 98306661-98306661 on Build GRCh38
rs780413594 Chr.7: 98306571-98306571 on Build GRCh38
rs779096013 Chr.7: 98306459-98306459 on Build GRCh38
rs772397097 Chr.7: 98306473-98306473 on Build GRCh38
rs747577265 Chr.7: 98306489-98306489 on Build GRCh38
rs768990780 Chr.7: 98306490-98306490 on Build GRCh38
rs749260503 Chr.7: 98306453-98306453 on Build GRCh38
rs778532601 Chr.7: 98306409-98306409 on Build GRCh38
rs1050490801 Chr.7: 98306303-98306303 on Build GRCh38
rs747273857 Chr.7: 98306574-98306574 on Build GRCh38
rs747731963 Chr.7: 98306598-98306598 on Build GRCh38
rs767352714 Chr.7: 98306545-98306545 on Build GRCh38
rs761212599 Chr.7: 98306496-98306496 on Build GRCh38
rs944779309 Chr.7: 98306617-98306617 on Build GRCh38
rs751577726 Chr.7: 98306445-98306445 on Build GRCh38
rs1056631482 Chr.7: 98306270-98306270 on Build GRCh38
rs1023991032 Chr.7: 98306386-98306386 on Build GRCh38
rs201317801 Chr.7: 98306427-98306427 on Build GRCh38
rs776267433 Chr.7: 98306495-98306495 on Build GRCh38
rs1049834062 Chr.7: 98306585-98306585 on Build GRCh38
rs757262879 Chr.7: 98306554-98306554 on Build GRCh38
rs189996945 Chr.7: 98306522-98306522 on Build GRCh38
rs996923271 Chr.7: 98306327-98306327 on Build GRCh38
rs898597526 Chr.7: 98306326-98306326 on Build GRCh38

View Full Product Details