The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127218.2 | NP_001120690.1 | 5 - 6 | 872 | 137 |
NM_001256879.1 | NP_001243808.1 | 6 - 7 | 1233 | 137 | |
NM_006230.3 | NP_006221.2 | 5 - 6 | 699 | 137 | |
XM_011515431.1 | XP_011513733.1 | 5 - 6 | 695 | 137 | |
GenBank mRNA | AK292347.1 | - | 6 - 7 | 1233 | 137 |
AK298168.1 | - | 5 - 6 | 563 | 137 | |
AK312294.1 | - | 5 - 6 | 832 | 137 | |
BC000459.2 | - | 5 - 6 | 662 | 137 | |
BC030001.2 | - | 4 - 5 | 639 | 137 | |
U21090.1 | - | 5 - 6 | 661 | 137 |