Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD4
Entrez Gene ID
54939
Gene Name
COMM domain containing 4
Gene Aliases
-
Chromosome Location
Chr.15: 75335996 - 75341927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284377.1 NP_001271306.1 4 - 5 279 139
NM_001284378.1 NP_001271307.1 4 - 5 279 139
NM_001284379.1 NP_001271308.1 3 - 4 257 139
NM_001321844.1 NP_001308773.1 4 - 5 273 139
NM_001321848.1 NP_001308777.1 3 - 4 257 139
NM_017828.4 NP_060298.2 4 - 5 279 139
NR_104312.1 4 - 5 279 139
XM_005254509.3 XP_005254566.1 4 - 5 305 139
XM_011521739.1 XP_011520041.1 4 - 5 314 139
XM_011521741.1 XP_011520043.1 4 - 5 314 139
GenBank mRNA AK000459.1 - 4 - 5 216 139
AK225716.1 - 4 - 5 240 139
AK314740.1 - 4 - 5 212 139
AL136872.1 - 4 - 5 199 139
AY542160.1 - 3 - 4 185 139
BC000837.2 - 3 - 4 182 139
BC068998.1 - 4 - 5 201 139
BI829264.1 - 4 - 5 255 139
BI829484.1 - 4 - 5 279 139
BQ233598.1 - - 242 139
CR533452.1 - 3 - 4 185 139
HY162014.1 - 3 - 4 258 139

View Full Product Details