The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303470.1 | NP_001290399.1 | 6 - 7 | 717 | 93 |
NM_018275.4 | NP_060745.3 | 6 - 7 | 1217 | 93 | |
GenBank mRNA | AK001787.1 | - | 2 - 3 | 197 | 93 |
AK054872.1 | - | 6 - 7 | 713 | 93 | |
AK302342.1 | - | 4 - 5 | 565 | 93 | |
BC002749.2 | - | 6 - 7 | 617 | 93 | |
BC015722.2 | - | 6 - 7 | 1175 | 93 |