Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PDP1
- Entrez Gene ID
-
54704
- Gene Name
- pyruvate dehyrogenase phosphatase catalytic subunit 1
- Gene Aliases
-
PDH, PDP, PDPC, PPM2A, PPM2C
- Chromosome Location
-
Chr.8: 93916855 - 93926068 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF155661.1 |
- |
1 - 2 |
92 |
90 |
View Full Product Details