The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018509.3 | NP_060979.2 | 6 - 7 | 914 | 101 |
GenBank mRNA | AF119857.1 | - | 3 - 4 | 272 | 101 |
AK316593.1 | - | 4 - 5 | 416 | 101 | |
BC017168.1 | - | 6 - 7 | 872 | 101 | |
BC052279.1 | - | 6 - 7 | 832 | 101 |