Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
Chromosome Location
Chr.17: 73193034 - 73208507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018714.2 NP_061184.1 10 - 11 2541 58
GenBank mRNA AB037802.1 - 10 - 11 2503 58
AK025633.1 - 7 - 8 1667 58
AK098224.1 - 10 - 11 2261 58
AL359611.1 - 4 - 5 881 58
BC021985.2 - 10 - 11 2506 58
BC047465.1 - 11 - 12 2843 58

View Full Product Details