The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243251.1 | NP_001230180.1 | 3 - 4 | 269 | 143 |
NM_024678.5 | NP_078954.4 | 3 - 4 | 750 | 143 | |
XM_011545253.1 | XP_011543555.1 | 3 - 4 | 743 | 143 | |
XM_017018302.1 | XP_016873791.1 | 3 - 4 | 743 | 143 | |
XM_017018303.1 | XP_016873792.1 | 3 - 4 | 262 | 143 | |
XM_017018304.1 | XP_016873793.1 | 3 - 4 | 262 | 143 | |
GenBank mRNA | AK056980.1 | - | 3 - 4 | 667 | 143 |
AK315425.1 | - | 3 - 4 | 256 | 143 | |
AW968557.1 | - | 3 - 4 | 467 | 143 | |
BC007800.2 | - | 3 - 4 | 560 | 143 | |
BP196275.1 | - | 3 - 4 | 269 | 143 |