Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
Chromosome Location
Chr.19: 8580240 - 8610735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282352.1 NP_001269281.1 1 - 2 243 73
NM_030957.3 NP_112219.3 14 - 15 2043 73
XM_006722917.3 XP_006722980.1 13 - 14 1889 73
XM_017027338.1 XP_016882827.1 13 - 14 1899 73
XM_017027339.1 XP_016882828.1 4 - 5 523 73
XM_017027340.1 XP_016882829.1 3 - 4 383 73
GenBank mRNA AB209515.1 - 13 - 14 1879 73
AK131414.1 - 1 - 2 243 73

View Full Product Details