The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032111.3 | NP_115487.2 | 1 - 2 | 147 | 74 |
GenBank mRNA | AK312086.1 | - | 1 - 2 | 79 | 74 |
BM559405.1 | - | - | 159 | 74 | |
BM910935.1 | - | - | 110 | 74 |