Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138076340-138076849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1D1
Entrez Gene ID
6718
Gene Name
aldo-keto reductase family 1, member D1
Gene Aliases
3o5bred, CBAS2, SRD5B1
UniGene
Hs.201667
Gene Chromosome Location
Chr.7: 138076432-138118304 on Build GRCh38
Transcripts
NM_001190906.1 NM_001190907.1 NM_005989.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309033 c.32C>G p.P11R Chr.7: 138076550-138076550 on Build GRCh38
6108985 c.80C>G p.S27* Chr.7: 138076598-138076598 on Build GRCh38
6597984 c.93+10delT p.? Chr.7: 138076621-138076621 on Build GRCh38
4828801 c.40G>A p.D14N Chr.7: 138076558-138076558 on Build GRCh38
5492316 c.24C>T p.H8H Chr.7: 138076542-138076542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046500465 Chr.7: 138076372-138076372 on Build GRCh38
rs895174386 Chr.7: 138076364-138076365 on Build GRCh38
rs768164623 Chr.7: 138076642-138076642 on Build GRCh38
rs776453053 Chr.7: 138076510-138076510 on Build GRCh38
rs779190099 Chr.7: 138076495-138076495 on Build GRCh38
rs1000519292 Chr.7: 138076446-138076446 on Build GRCh38
rs949459070 Chr.7: 138076365-138076365 on Build GRCh38
rs780692011 Chr.7: 138076581-138076581 on Build GRCh38
rs780985612 Chr.7: 138076590-138076590 on Build GRCh38
rs373481986 Chr.7: 138076485-138076485 on Build GRCh38
rs745652944 Chr.7: 138076592-138076592 on Build GRCh38
rs925181907 Chr.7: 138076664-138076664 on Build GRCh38
rs999132653 Chr.7: 138076425-138076425 on Build GRCh38
rs145509975 Chr.7: 138076473-138076473 on Build GRCh38
rs148565883 Chr.7: 138076363-138076363 on Build GRCh38
rs746817839 Chr.7: 138076496-138076496 on Build GRCh38
rs794726925 Chr.7: 138076528-138076528 on Build GRCh38
rs780215526 Chr.7: 138076487-138076487 on Build GRCh38
rs545899980 Chr.7: 138076665-138076665 on Build GRCh38
rs893069921 Chr.7: 138076434-138076434 on Build GRCh38
rs757029846 Chr.7: 138076588-138076588 on Build GRCh38
rs7799469 Chr.7: 138076622-138076622 on Build GRCh38
rs750969279 Chr.7: 138076576-138076576 on Build GRCh38
rs201065248 Chr.7: 138076582-138076582 on Build GRCh38
rs779049914 Chr.7: 138076585-138076585 on Build GRCh38
rs755075482 Chr.7: 138076584-138076584 on Build GRCh38
rs113046960 Chr.7: 138076440-138076440 on Build GRCh38
rs926576419 Chr.7: 138076553-138076553 on Build GRCh38
rs776924075 Chr.7: 138076648-138076652 on Build GRCh38
rs199742311 Chr.7: 138076632-138076632 on Build GRCh38
rs1030089892 Chr.7: 138076494-138076494 on Build GRCh38
rs748348035 Chr.7: 138076586-138076586 on Build GRCh38
rs554884607 Chr.7: 138076445-138076445 on Build GRCh38
rs886214809 Chr.7: 138076783-138076783 on Build GRCh38
rs907036941 Chr.7: 138076394-138076394 on Build GRCh38
rs377690951 Chr.7: 138076659-138076659 on Build GRCh38
rs759374141 Chr.7: 138076543-138076543 on Build GRCh38
rs900419297 Chr.7: 138076800-138076800 on Build GRCh38
rs751204230 Chr.7: 138076563-138076563 on Build GRCh38
rs900133884 Chr.7: 138076703-138076703 on Build GRCh38
rs761463498 Chr.7: 138076658-138076658 on Build GRCh38
rs769791635 Chr.7: 138076601-138076601 on Build GRCh38
rs756630815 Chr.7: 138076578-138076578 on Build GRCh38
rs759406658 Chr.7: 138076534-138076534 on Build GRCh38
rs1053384383 Chr.7: 138076663-138076663 on Build GRCh38
rs190907624 Chr.7: 138076573-138076573 on Build GRCh38
rs749533931 Chr.7: 138076488-138076488 on Build GRCh38
rs7780438 Chr.7: 138076504-138076504 on Build GRCh38
rs374278061 Chr.7: 138076645-138076645 on Build GRCh38
rs767977652 Chr.7: 138076641-138076644 on Build GRCh38
rs138462322 Chr.7: 138076542-138076542 on Build GRCh38
rs148732404 Chr.7: 138076610-138076610 on Build GRCh38
rs534954215 Chr.7: 138076441-138076441 on Build GRCh38
rs773770019 Chr.7: 138076661-138076661 on Build GRCh38
rs143960528 Chr.7: 138076567-138076567 on Build GRCh38
rs748847040 Chr.7: 138076617-138076617 on Build GRCh38
rs757985744 Chr.7: 138076432-138076432 on Build GRCh38
rs537010232 Chr.7: 138076544-138076544 on Build GRCh38
rs763696096 Chr.7: 138076547-138076547 on Build GRCh38
rs771432374 Chr.7: 138076633-138076636 on Build GRCh38
rs933014722 Chr.7: 138076753-138076753 on Build GRCh38
rs945202972 Chr.7: 138076395-138076395 on Build GRCh38
rs7799169 Chr.7: 138076464-138076464 on Build GRCh38
rs374681767 Chr.7: 138076493-138076493 on Build GRCh38
rs770734480 Chr.7: 138076508-138076508 on Build GRCh38
rs1052947797 Chr.7: 138076429-138076429 on Build GRCh38
rs1038932109 Chr.7: 138076789-138076789 on Build GRCh38
rs769312349 Chr.7: 138076539-138076539 on Build GRCh38
rs1004359528 Chr.7: 138076509-138076509 on Build GRCh38
rs936611576 Chr.7: 138076670-138076670 on Build GRCh38
rs545579608 Chr.7: 138076668-138076668 on Build GRCh38
rs781171275 Chr.7: 138076799-138076799 on Build GRCh38
rs59586431 Chr.7: 138076673-138076673 on Build GRCh38
rs916587979 Chr.7: 138076612-138076612 on Build GRCh38
rs760874257 Chr.7: 138076808-138076808 on Build GRCh38
rs900247975 Chr.7: 138076480-138076480 on Build GRCh38

View Full Product Details