Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138834941-138835455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1549
Entrez Gene ID
57670
Gene Name
KIAA1549
Gene Aliases
-
UniGene
Hs.605380
Gene Chromosome Location
Chr.7: 138831377-138981626 on Build GRCh38
Transcripts
NM_001164665.1 NM_020910.2 XM_011516442.2

SNP Detail

SNP ID Chromosome Location
rs919023204 Chr.7: 138835345-138835345 on Build GRCh38
rs150862041 Chr.7: 138835211-138835212 on Build GRCh38
rs1040444235 Chr.7: 138835263-138835266 on Build GRCh38
rs559899581 Chr.7: 138835370-138835370 on Build GRCh38
rs1047576213 Chr.7: 138835297-138835297 on Build GRCh38
rs1006074907 Chr.7: 138835142-138835142 on Build GRCh38
rs191637263 Chr.7: 138834990-138834990 on Build GRCh38
rs377173445 Chr.7: 138835203-138835203 on Build GRCh38
rs950211714 Chr.7: 138835034-138835034 on Build GRCh38
rs879404761 Chr.7: 138835383-138835383 on Build GRCh38
rs963040617 Chr.7: 138835332-138835332 on Build GRCh38
rs1026151996 Chr.7: 138834973-138834973 on Build GRCh38
rs918991889 Chr.7: 138835105-138835105 on Build GRCh38
rs565063232 Chr.7: 138834973-138834973 on Build GRCh38
rs73458272 Chr.7: 138835319-138835319 on Build GRCh38
rs1024353707 Chr.7: 138835186-138835186 on Build GRCh38
rs1006104648 Chr.7: 138835317-138835317 on Build GRCh38
rs189796953 Chr.7: 138835257-138835257 on Build GRCh38
rs183040202 Chr.7: 138835123-138835123 on Build GRCh38
rs561677583 Chr.7: 138835131-138835131 on Build GRCh38
rs1039000659 Chr.7: 138835409-138835409 on Build GRCh38
rs984122980 Chr.7: 138834984-138834986 on Build GRCh38
rs779427430 Chr.7: 138835058-138835058 on Build GRCh38
rs541469040 Chr.7: 138835126-138835126 on Build GRCh38
rs398006559 Chr.7: 138835217-138835218 on Build GRCh38
rs1017367387 Chr.7: 138835159-138835159 on Build GRCh38
rs910218780 Chr.7: 138835236-138835236 on Build GRCh38
rs991564007 Chr.7: 138835018-138835019 on Build GRCh38
rs911770904 Chr.7: 138835363-138835363 on Build GRCh38
rs1054147272 Chr.7: 138835066-138835066 on Build GRCh38
rs987714644 Chr.7: 138835348-138835348 on Build GRCh38
rs575127650 Chr.7: 138835182-138835182 on Build GRCh38
rs186365916 Chr.7: 138835230-138835230 on Build GRCh38
rs973158738 Chr.7: 138835104-138835104 on Build GRCh38
rs772024198 Chr.7: 138835188-138835188 on Build GRCh38
rs552671651 Chr.7: 138835106-138835106 on Build GRCh38
rs959066073 Chr.7: 138835015-138835015 on Build GRCh38
rs11767272 Chr.7: 138835077-138835077 on Build GRCh38
rs775682534 Chr.7: 138835197-138835197 on Build GRCh38
rs970717642 Chr.7: 138835291-138835291 on Build GRCh38
rs768984088 Chr.7: 138835398-138835398 on Build GRCh38
rs887586854 Chr.7: 138835304-138835304 on Build GRCh38
rs958702104 Chr.7: 138835175-138835175 on Build GRCh38
rs924895565 Chr.7: 138835059-138835059 on Build GRCh38
rs1051580324 Chr.7: 138835122-138835122 on Build GRCh38
rs891863129 Chr.7: 138835253-138835253 on Build GRCh38
rs1000184979 Chr.7: 138835193-138835193 on Build GRCh38
rs544134873 Chr.7: 138835191-138835191 on Build GRCh38
rs1017133016 Chr.7: 138835322-138835322 on Build GRCh38
rs201578676 Chr.7: 138835180-138835180 on Build GRCh38
rs917416615 Chr.7: 138835024-138835025 on Build GRCh38
rs971468164 Chr.7: 138835192-138835192 on Build GRCh38
rs200602963 Chr.7: 138834986-138834986 on Build GRCh38
rs1039363473 Chr.7: 138835132-138835132 on Build GRCh38
rs909871353 Chr.7: 138834984-138834984 on Build GRCh38
rs397791719 Chr.7: 138834985-138834986 on Build GRCh38
rs951963726 Chr.7: 138834983-138834984 on Build GRCh38
rs1041994272 Chr.7: 138835035-138835035 on Build GRCh38
rs901628455 Chr.7: 138835281-138835281 on Build GRCh38
rs539226063 Chr.7: 138834983-138834983 on Build GRCh38
rs386718364 Chr.7: 138835181-138835188 on Build GRCh38
rs936230305 Chr.7: 138835062-138835062 on Build GRCh38
rs200471820 Chr.7: 138835185-138835188 on Build GRCh38
rs150473976 Chr.7: 138835094-138835094 on Build GRCh38
rs879928106 Chr.7: 138835333-138835333 on Build GRCh38
rs950465870 Chr.7: 138835346-138835346 on Build GRCh38
rs767904035 Chr.7: 138834972-138834973 on Build GRCh38
rs1014851153 Chr.7: 138835254-138835254 on Build GRCh38
rs973021872 Chr.7: 138835335-138835335 on Build GRCh38
rs71169063 Chr.7: 138835212-138835213 on Build GRCh38
rs963011592 Chr.7: 138835102-138835102 on Build GRCh38
rs980852787 Chr.7: 138835027-138835027 on Build GRCh38

View Full Product Details