Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138836857-138837352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1549
Entrez Gene ID
57670
Gene Name
KIAA1549
Gene Aliases
-
UniGene
Hs.605380
Gene Chromosome Location
Chr.7: 138831377-138981626 on Build GRCh38
Transcripts
NM_001164665.1 NM_020910.2 XM_011516442.2

SNP Detail

SNP ID Chromosome Location
rs956438694 Chr.7: 138837182-138837182 on Build GRCh38
rs907061013 Chr.7: 138836984-138836984 on Build GRCh38
rs1004198398 Chr.7: 138837307-138837307 on Build GRCh38
rs1020134870 Chr.7: 138837326-138837326 on Build GRCh38
rs964328598 Chr.7: 138837244-138837244 on Build GRCh38
rs774352738 Chr.7: 138836991-138836991 on Build GRCh38
rs555635640 Chr.7: 138837293-138837293 on Build GRCh38
rs774795710 Chr.7: 138837137-138837137 on Build GRCh38
rs936871665 Chr.7: 138837142-138837142 on Build GRCh38
rs575429012 Chr.7: 138837298-138837298 on Build GRCh38
rs921936340 Chr.7: 138837039-138837039 on Build GRCh38
rs112222953 Chr.7: 138837181-138837182 on Build GRCh38
rs527548363 Chr.7: 138837165-138837165 on Build GRCh38
rs1041883653 Chr.7: 138837180-138837180 on Build GRCh38
rs995475597 Chr.7: 138837014-138837014 on Build GRCh38
rs148657552 Chr.7: 138837120-138837120 on Build GRCh38
rs375726770 Chr.7: 138836937-138836937 on Build GRCh38
rs546907841 Chr.7: 138836895-138836895 on Build GRCh38
rs1010360382 Chr.7: 138836980-138836980 on Build GRCh38
rs908053691 Chr.7: 138837085-138837085 on Build GRCh38
rs368544875 Chr.7: 138837011-138837011 on Build GRCh38
rs568941491 Chr.7: 138837026-138837026 on Build GRCh38
rs921497406 Chr.7: 138836883-138836883 on Build GRCh38
rs759377606 Chr.7: 138837027-138837027 on Build GRCh38
rs965952750 Chr.7: 138837115-138837115 on Build GRCh38
rs1037866375 Chr.7: 138837112-138837112 on Build GRCh38
rs151210817 Chr.7: 138837242-138837242 on Build GRCh38
rs766192333 Chr.7: 138836881-138836882 on Build GRCh38
rs1030854318 Chr.7: 138836980-138836983 on Build GRCh38
rs74351257 Chr.7: 138837069-138837069 on Build GRCh38
rs970909007 Chr.7: 138836911-138836911 on Build GRCh38
rs533751517 Chr.7: 138837174-138837174 on Build GRCh38
rs894400382 Chr.7: 138837250-138837250 on Build GRCh38
rs17160567 Chr.7: 138836880-138836880 on Build GRCh38
rs921525521 Chr.7: 138837127-138837127 on Build GRCh38
rs1010777027 Chr.7: 138837219-138837219 on Build GRCh38
rs193045454 Chr.7: 138837271-138837271 on Build GRCh38
rs1034250534 Chr.7: 138836992-138836992 on Build GRCh38
rs571301467 Chr.7: 138837167-138837167 on Build GRCh38
rs1026904779 Chr.7: 138837022-138837022 on Build GRCh38
rs767503474 Chr.7: 138837101-138837101 on Build GRCh38
rs763606115 Chr.7: 138837193-138837193 on Build GRCh38
rs975177008 Chr.7: 138837012-138837012 on Build GRCh38
rs142173573 Chr.7: 138837227-138837227 on Build GRCh38
rs1003304377 Chr.7: 138837168-138837168 on Build GRCh38
rs566586016 Chr.7: 138836894-138836894 on Build GRCh38
rs533145548 Chr.7: 138836893-138836893 on Build GRCh38
rs765583971 Chr.7: 138837308-138837308 on Build GRCh38
rs770816753 Chr.7: 138836961-138836961 on Build GRCh38
rs544631263 Chr.7: 138836982-138836982 on Build GRCh38
rs1026852261 Chr.7: 138837272-138837272 on Build GRCh38

View Full Product Details