The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170686.1 | NP_001164157.1 | 3 - 4 | 639 | 65 |
NM_001170687.1 | NP_001164158.1 | 3 - 4 | 597 | 65 | |
NM_001170688.1 | NP_001164159.1 | 3 - 4 | 639 | 65 | |
NM_001170689.1 | NP_001164160.1 | 3 - 4 | 614 | 65 | |
NM_080875.2 | NP_543151.2 | 3 - 4 | 639 | 65 | |
XM_006710372.1 | XP_006710435.1 | 3 - 4 | 611 | 65 | |
XM_017000358.1 | XP_016855847.1 | 3 - 4 | 611 | 65 | |
XM_017000359.1 | XP_016855848.1 | 3 - 4 | 611 | 65 | |
XM_017000360.1 | XP_016855849.1 | 3 - 4 | 558 | 65 | |
XM_017000361.1 | XP_016855850.1 | 3 - 4 | 611 | 65 | |
XM_017000362.1 | XP_016855851.1 | 3 - 4 | 651 | 65 | |
XM_017000363.1 | XP_016855852.1 | 3 - 4 | 614 | 65 | |
XM_017000364.1 | XP_016855853.1 | 3 - 4 | 611 | 65 | |
GenBank mRNA | AB064367.1 | - | 1 - 2 | 46 | 65 |
AB074480.1 | - | 3 - 4 | 550 | 65 | |
AB076691.1 | - | 3 - 4 | 508 | 65 | |
AB076692.1 | - | 3 - 4 | 550 | 65 | |
AB076693.1 | - | 3 - 4 | 550 | 65 | |
AB097000.1 | - | 3 - 4 | 473 | 65 | |
AK098785.1 | - | 3 - 4 | 508 | 65 | |
AK122863.1 | - | 3 - 4 | 540 | 65 | |
AK128167.1 | - | 3 - 4 | 574 | 65 | |
AK128553.1 | - | 3 - 4 | 558 | 65 | |
AK293642.1 | - | 2 - 3 | 619 | 65 | |
AK302757.1 | - | 3 - 4 | 591 | 65 | |
AK309362.1 | - | 2 - 3 | 617 | 65 | |
BC016490.1 | - | 3 - 4 | 476 | 65 | |
BC037542.1 | - | 3 - 4 | 519 | 65 |