Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNG3
Entrez Gene ID
170850
Gene Name
potassium voltage-gated channel modifier subfamily G member 3
Gene Aliases
KV10.1, KV6.3
Chromosome Location
Chr.2: 42442017 - 42494097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133329.5 NP_579875.1 1 - 2 1262 83
GenBank mRNA AB070604.1 - 1 - 2 666 83
AF348982.1 - 1 - 2 1261 83
AF454548.1 - 1 - 2 1143 83
BC071558.1 - 1 - 2 1142 83

View Full Product Details