Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141621346-141621849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGK
Entrez Gene ID
55750
Gene Name
acylglycerol kinase
Gene Aliases
CATC5, CTRCT38, MTDPS10, MULK
UniGene
Hs.743318
Gene Chromosome Location
Chr.7: 141551278-141655244 on Build GRCh38
Transcripts
NM_018238.3 XM_005250023.3 XM_011516397.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1568582 c.585C>A p.I195I Chr.7: 141621798-141621798 on Build GRCh38
1330033 c.556G>C p.E186Q Chr.7: 141621769-141621769 on Build GRCh38
3662983 c.588+7C>G p.? Chr.7: 141621808-141621808 on Build GRCh38
4978086 c.562G>A p.V188I Chr.7: 141621775-141621775 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003157595 Chr.7: 141621709-141621709 on Build GRCh38
rs747244579 Chr.7: 141621775-141621775 on Build GRCh38
rs751795898 Chr.7: 141621407-141621407 on Build GRCh38
rs142576028 Chr.7: 141621635-141621636 on Build GRCh38
rs953177561 Chr.7: 141621707-141621707 on Build GRCh38
rs1014813140 Chr.7: 141621614-141621623 on Build GRCh38
rs540365139 Chr.7: 141621543-141621543 on Build GRCh38
rs200104400 Chr.7: 141621757-141621757 on Build GRCh38
rs1004812805 Chr.7: 141621611-141621611 on Build GRCh38
rs41275000 Chr.7: 141621448-141621448 on Build GRCh38
rs759819550 Chr.7: 141621707-141621709 on Build GRCh38
rs1042123231 Chr.7: 141621628-141621628 on Build GRCh38
rs748709307 Chr.7: 141621750-141621750 on Build GRCh38
rs997382936 Chr.7: 141621634-141621635 on Build GRCh38
rs953004378 Chr.7: 141621480-141621480 on Build GRCh38
rs367778952 Chr.7: 141621816-141621816 on Build GRCh38
rs770005760 Chr.7: 141621726-141621727 on Build GRCh38
rs528072860 Chr.7: 141621503-141621503 on Build GRCh38
rs367592307 Chr.7: 141621663-141621663 on Build GRCh38
rs755469922 Chr.7: 141621727-141621727 on Build GRCh38
rs1052607111 Chr.7: 141621535-141621535 on Build GRCh38
rs78925883 Chr.7: 141621608-141621608 on Build GRCh38
rs1029228849 Chr.7: 141621699-141621699 on Build GRCh38
rs1035903385 Chr.7: 141621434-141621434 on Build GRCh38
rs922600853 Chr.7: 141621570-141621570 on Build GRCh38
rs995980072 Chr.7: 141621591-141621592 on Build GRCh38
rs902303518 Chr.7: 141621667-141621667 on Build GRCh38
rs150053901 Chr.7: 141621596-141621597 on Build GRCh38
rs965951700 Chr.7: 141621621-141621621 on Build GRCh38
rs148583259 Chr.7: 141621554-141621554 on Build GRCh38
rs893615320 Chr.7: 141621616-141621616 on Build GRCh38
rs918093334 Chr.7: 141621540-141621540 on Build GRCh38
rs547375746 Chr.7: 141621516-141621518 on Build GRCh38
rs887080312 Chr.7: 141621596-141621599 on Build GRCh38
rs776709096 Chr.7: 141621786-141621786 on Build GRCh38
rs1045633823 Chr.7: 141621547-141621547 on Build GRCh38
rs908642732 Chr.7: 141621523-141621523 on Build GRCh38
rs1048410055 Chr.7: 141621594-141621607 on Build GRCh38
rs778353485 Chr.7: 141621766-141621766 on Build GRCh38
rs1013146663 Chr.7: 141621759-141621759 on Build GRCh38
rs916055520 Chr.7: 141621453-141621453 on Build GRCh38
rs202037926 Chr.7: 141621790-141621790 on Build GRCh38
rs980773447 Chr.7: 141621578-141621603 on Build GRCh38
rs988354902 Chr.7: 141621822-141621822 on Build GRCh38
rs369065604 Chr.7: 141621627-141621627 on Build GRCh38
rs1050120909 Chr.7: 141621597-141621597 on Build GRCh38
rs748227619 Chr.7: 141621478-141621478 on Build GRCh38
rs924921087 Chr.7: 141621464-141621464 on Build GRCh38
rs753290543 Chr.7: 141621697-141621697 on Build GRCh38
rs374793939 Chr.7: 141621712-141621712 on Build GRCh38
rs114181612 Chr.7: 141621437-141621437 on Build GRCh38
rs771075292 Chr.7: 141621779-141621779 on Build GRCh38
rs946564524 Chr.7: 141621618-141621618 on Build GRCh38
rs1034616657 Chr.7: 141621747-141621747 on Build GRCh38
rs532781515 Chr.7: 141621599-141621599 on Build GRCh38
rs1003188558 Chr.7: 141621411-141621411 on Build GRCh38
rs932522307 Chr.7: 141621582-141621582 on Build GRCh38
rs745967065 Chr.7: 141621792-141621792 on Build GRCh38
rs863223896 Chr.7: 141621769-141621769 on Build GRCh38
rs779581356 Chr.7: 141621748-141621748 on Build GRCh38

View Full Product Details