Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN3
Entrez Gene ID
9369
Gene Name
neurexin 3
Gene Aliases
C14orf60
Chromosome Location
Chr.14: 77979904 - 79868290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105250.2 NP_001098720.1 5 - 6 1953 105
NM_001272020.1 NP_001258949.1 4 - 5 1863 105
NM_004796.5 NP_004787.2 15 - 16 3316 105
NM_138970.4 NP_620426.2 4 - 5 1863 105
NR_073546.1 5 - 6 1953 105
NR_073547.1 18 - 19 4987 105
XM_005268218.2 XP_005268275.1 21 - 22 5056 105
XM_006720322.2 XP_006720385.1 20 - 21 5044 105
XM_006720323.2 XP_006720386.1 19 - 20 5020 105
XM_011537363.1 XP_011535665.1 21 - 22 5056 105
XM_011537364.2 XP_011535666.1 22 - 23 5018 105
XM_011537365.2 XP_011535667.1 21 - 22 4844 105
XM_011537366.1 XP_011535668.1 20 - 21 5032 105
XM_011537367.1 XP_011535669.1 20 - 21 4966 105
XM_011537368.1 XP_011535670.1 21 - 22 5056 105
XM_011537369.1 XP_011535671.1 21 - 22 5056 105
XM_011537370.1 XP_011535672.1 20 - 21 5032 105
XM_011537371.1 XP_011535673.1 21 - 22 5056 105
XM_011537372.1 XP_011535674.1 21 - 22 5056 105
XM_011537373.1 XP_011535675.1 20 - 21 4966 105
XM_011537377.2 XP_011535679.1 17 - 18 3348 105
XM_017021790.1 XP_016877279.1 22 - 23 5156 105
XM_017021791.1 XP_016877280.1 22 - 23 4939 105
XM_017021792.1 XP_016877281.1 20 - 21 4839 105
XM_017021793.1 XP_016877282.1 20 - 21 5029 105
XM_017021794.1 XP_016877283.1 19 - 20 5020 105
XM_017021795.1 XP_016877284.1 19 - 20 4815 105
XM_017021796.1 XP_016877285.1 19 - 20 5005 105
XM_017021797.1 XP_016877286.1 18 - 19 4996 105
XM_017021798.1 XP_016877287.1 18 - 19 4993 105
XM_017021799.1 XP_016877288.1 20 - 21 4966 105
XM_017021800.1 XP_016877289.1 20 - 21 4966 105
XM_017021801.1 XP_016877290.1 19 - 20 4939 105
XM_017021802.1 XP_016877291.1 18 - 19 4960 105
XM_017021803.1 XP_016877292.1 20 - 21 5035 105
XM_017021804.1 XP_016877293.1 20 - 21 5029 105
XM_017021805.1 XP_016877294.1 19 - 20 5020 105
XM_017021806.1 XP_016877295.1 19 - 20 5005 105
XM_017021807.1 XP_016877296.1 20 - 21 4966 105
GenBank mRNA AB018286.1 - 15 - 16 3297 105
AJ316284.2 - 18 - 19 4775 105
AJ493127.1 - 5 - 6 1029 105
AK056530.1 - 4 - 5 1863 105
AL833739.1 - 4 - 5 1158 105
BC059368.1 - 4 - 5 1789 105
BC068469.1 - 4 - 5 1789 105
BC142649.1 - 5 - 6 1386 105
BC150194.1 - 5 - 6 1386 105
BC152457.1 - 15 - 16 3297 105

View Full Product Details