The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004160.5 | NP_004151.3 | 6 - 7 | 812 | 150 |
XM_011525035.1 | XP_011523337.1 | 6 - 7 | 1422 | 150 | |
GenBank mRNA | BC041057.1 | - | 6 - 7 | 812 | 150 |
D13899.1 | - | 3 - 4 | 351 | 150 |