Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38411421-38411945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
UniGene
Hs.264887
Gene Chromosome Location
Chr.8: 38411138-38468834 on Build GRCh38
Transcripts
NM_001174063.1 NM_001174064.1 NM_001174065.1 NM_001174066.1 NM_001174067.1 NM_015850.3 NM_023105.2 NM_023106.2 NM_023110.2 XM_006716303.2 XM_006716304.1 XM_006716306.2 XM_006716307.1 XM_006716309.3 XM_006716310.2 XM_006716311.1 XM_006716312.1 XM_006716313.2 XM_006716314.1 XM_011544443.2 XM_011544444.1 XM_011544445.2 XM_011544446.2 XM_011544447.2 XM_011544448.1 XM_011544449.1 XM_011544450.2 XM_011544451.1 XM_017013219.1 XM_017013220.1 XM_017013221.1 XM_017013222.1 XM_017013223.1 XM_017013224.1 XM_017013225.1 XM_017013226.1 XM_017013227.1 XM_017013228.1 XM_017013229.1 XM_017013230.1

SNP Detail

SNP ID Chromosome Location
rs35229167 Chr.8: 38411820-38411821 on Build GRCh38
rs868119571 Chr.8: 38411783-38411783 on Build GRCh38
rs539815580 Chr.8: 38411698-38411698 on Build GRCh38
rs981514968 Chr.8: 38411697-38411697 on Build GRCh38
rs892952105 Chr.8: 38411742-38411742 on Build GRCh38
rs886062907 Chr.8: 38411663-38411663 on Build GRCh38
rs1055136025 Chr.8: 38411904-38411904 on Build GRCh38
rs546856626 Chr.8: 38411468-38411468 on Build GRCh38
rs974305182 Chr.8: 38411659-38411659 on Build GRCh38
rs902383810 Chr.8: 38411737-38411737 on Build GRCh38
rs567642271 Chr.8: 38411917-38411917 on Build GRCh38
rs939397751 Chr.8: 38411877-38411877 on Build GRCh38
rs1055506238 Chr.8: 38411717-38411720 on Build GRCh38
rs962901340 Chr.8: 38411636-38411636 on Build GRCh38
rs577559730 Chr.8: 38411507-38411508 on Build GRCh38
rs937098717 Chr.8: 38411722-38411722 on Build GRCh38
rs929652203 Chr.8: 38411694-38411694 on Build GRCh38
rs940045661 Chr.8: 38411475-38411475 on Build GRCh38
rs528523076 Chr.8: 38411451-38411451 on Build GRCh38
rs906565559 Chr.8: 38411503-38411503 on Build GRCh38
rs542519157 Chr.8: 38411792-38411792 on Build GRCh38
rs910120036 Chr.8: 38411847-38411847 on Build GRCh38
rs962870329 Chr.8: 38411837-38411837 on Build GRCh38
rs776304980 Chr.8: 38411717-38411717 on Build GRCh38
rs893025397 Chr.8: 38411525-38411525 on Build GRCh38
rs16887356 Chr.8: 38411529-38411529 on Build GRCh38
rs143775607 Chr.8: 38411556-38411556 on Build GRCh38
rs966158446 Chr.8: 38411785-38411785 on Build GRCh38
rs188827615 Chr.8: 38411596-38411596 on Build GRCh38
rs570038633 Chr.8: 38411806-38411806 on Build GRCh38
rs768193115 Chr.8: 38411693-38411693 on Build GRCh38
rs918884611 Chr.8: 38411666-38411666 on Build GRCh38
rs574988190 Chr.8: 38411784-38411784 on Build GRCh38
rs549566640 Chr.8: 38411859-38411859 on Build GRCh38
rs556829066 Chr.8: 38411571-38411571 on Build GRCh38
rs551625170 Chr.8: 38411526-38411526 on Build GRCh38
rs999969013 Chr.8: 38411570-38411570 on Build GRCh38
rs891183872 Chr.8: 38411794-38411794 on Build GRCh38
rs928083466 Chr.8: 38411865-38411865 on Build GRCh38
rs371446312 Chr.8: 38411473-38411473 on Build GRCh38
rs1056050085 Chr.8: 38411521-38411521 on Build GRCh38
rs999297539 Chr.8: 38411777-38411777 on Build GRCh38
rs906982605 Chr.8: 38411484-38411484 on Build GRCh38
rs1003939301 Chr.8: 38411505-38411506 on Build GRCh38
rs572424562 Chr.8: 38411794-38411795 on Build GRCh38
rs771021553 Chr.8: 38411592-38411592 on Build GRCh38
rs955646941 Chr.8: 38411576-38411576 on Build GRCh38
rs1011200259 Chr.8: 38411718-38411718 on Build GRCh38
rs191921266 Chr.8: 38411729-38411729 on Build GRCh38
rs546296836 Chr.8: 38411840-38411840 on Build GRCh38
rs940161714 Chr.8: 38411673-38411673 on Build GRCh38
rs910088749 Chr.8: 38411660-38411660 on Build GRCh38
rs1001334345 Chr.8: 38411511-38411511 on Build GRCh38
rs372106811 Chr.8: 38411510-38411511 on Build GRCh38
rs916602331 Chr.8: 38411915-38411915 on Build GRCh38
rs1010752002 Chr.8: 38411550-38411550 on Build GRCh38
rs183394116 Chr.8: 38411858-38411858 on Build GRCh38
rs751945489 Chr.8: 38411591-38411592 on Build GRCh38
rs1041608851 Chr.8: 38411776-38411776 on Build GRCh38
rs536000259 Chr.8: 38411646-38411646 on Build GRCh38
rs947260762 Chr.8: 38411773-38411773 on Build GRCh38
rs563863786 Chr.8: 38411800-38411800 on Build GRCh38
rs984294403 Chr.8: 38411647-38411647 on Build GRCh38
rs183426730 Chr.8: 38411587-38411587 on Build GRCh38
rs999494508 Chr.8: 38411791-38411791 on Build GRCh38
rs201364530 Chr.8: 38411524-38411524 on Build GRCh38
rs572492442 Chr.8: 38411682-38411682 on Build GRCh38

View Full Product Details