Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE1A
Entrez Gene ID
5136
Gene Name
phosphodiesterase 1A
Gene Aliases
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A
Chromosome Location
Chr.2: 182140035 - 182523375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003683.2 NP_001003683.1 7 - 8 916 98
NM_001258312.1 NP_001245241.1 7 - 8 943 98
NM_001258313.1 NP_001245242.1 6 - 7 1095 98
NM_001258314.1 NP_001245243.1 5 - 6 772 98
NM_005019.4 NP_005010.2 6 - 7 958 98
XM_011511323.2 XP_011509625.1 6 - 7 735 98
XM_011511324.2 XP_011509626.1 7 - 8 963 98
XM_011511325.2 XP_011509627.1 6 - 7 1090 98
XM_011511326.2 XP_011509628.1 5 - 6 767 98
XM_017004294.1 XP_016859783.1 6 - 7 958 98
XM_017004295.1 XP_016859784.1 7 - 8 963 98
XM_017004296.1 XP_016859785.1 7 - 8 885 98
XM_017004297.1 XP_016859786.1 7 - 8 826 98
XM_017004298.1 XP_016859787.1 7 - 8 1008 98
XM_017004299.1 XP_016859788.1 7 - 8 963 98
XM_017004300.1 XP_016859789.1 7 - 8 1008 98
XM_017004301.1 XP_016859790.1 7 - 8 813 98
XM_017004302.1 XP_016859791.1 6 - 7 1090 98
GenBank mRNA AJ401610.1 - 6 - 7 677 98
AK294239.1 - 7 - 8 878 98
AK295657.1 - 7 - 8 748 98
AK301720.1 - 5 - 6 755 98
AL110263.1 - 6 - 7 1069 98
BC022480.1 - 6 - 7 926 98
U40370.1 - 7 - 8 809 98

View Full Product Details