Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BOP1
Entrez Gene ID
23246
Gene Name
block of proliferation 1
Gene Aliases
-
Chromosome Location
Chr.8: 144262046 - 144291438 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015201.4 NP_056016.1 10 - 11 1356 109
GenBank mRNA AB060694.1 - 9 - 10 1024 109
AK024840.1 - 10 - 11 1329 109
AK290677.1 - 10 - 11 1318 109
AK293719.1 - 9 - 10 1000 109
AK310015.1 - 8 - 9 930 109
AK315612.1 - 10 - 11 1325 109
BC001086.2 - 9 - 10 1082 109
BC005160.2 - 9 - 10 1109 109
BC007274.1 - 10 - 11 1251 109
BC013787.1 - 10 - 11 1318 109
BC013980.2 - 10 - 11 1321 109
BC017674.2 - 10 - 11 1308 109
BC073813.1 - 5 - 6 615 109
BC098446.1 - 6 - 7 752 109
D50914.1 - 9 - 10 1096 109

View Full Product Details