Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D10B
Entrez Gene ID
26000
Gene Name
TBC1 domain family member 10B
Gene Aliases
EPI64B, FP2461
Chromosome Location
Chr.16: 30357101 - 30370258 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015527.3 NP_056342.3 2 - 3 1068 126
XM_011545790.2 XP_011544092.1 2 - 3 1125 126
GenBank mRNA AB449895.1 - 2 - 3 585 126
AK023192.1 - 2 - 3 447 126
BC063112.1 - 2 - 3 492 126
BC072453.1 - 2 - 3 380 126
BC093814.1 - 2 - 3 296 126
BC093816.1 - 2 - 3 296 126
BC098419.1 - 2 - 3 632 126

View Full Product Details