Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
Chromosome Location
Chr.17: 60677811 - 61392838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099432.1 NP_001092902.1 7 - 8 591 82
NM_001320470.1 NP_001307399.1 7 - 8 591 82
NM_017679.3 NP_060149.3 7 - 8 591 82
XM_005257472.1 XP_005257529.1 7 - 8 542 82
XM_011524939.1 XP_011523241.1 7 - 8 542 82
XM_011524940.1 XP_011523242.1 7 - 8 542 82
XM_011524941.1 XP_011523243.1 7 - 8 542 82
XM_011524942.2 XP_011523244.1 7 - 8 542 82
XM_011524943.2 XP_011523245.1 7 - 8 542 82
XM_011524944.1 XP_011523246.1 7 - 8 542 82
XM_011524945.2 XP_011523247.1 7 - 8 542 82
XM_011524946.2 XP_011523248.1 7 - 8 542 82
XM_011524947.1 XP_011523249.1 7 - 8 542 82
XM_011524948.2 XP_011523250.1 7 - 8 542 82
XM_011524949.2 XP_011523251.1 7 - 8 542 82
XM_011524950.1 XP_011523252.1 7 - 8 542 82
XM_011524951.2 XP_011523253.1 7 - 8 542 82
XM_017024783.1 XP_016880272.1 7 - 8 542 82
XM_017024784.1 XP_016880273.1 7 - 8 542 82
XM_017024785.1 XP_016880274.1 7 - 8 542 82
XM_017024786.1 XP_016880275.1 7 - 8 542 82
XM_017024787.1 XP_016880276.1 7 - 8 542 82
XM_017024788.1 XP_016880277.1 7 - 8 542 82
XM_017024789.1 XP_016880278.1 7 - 8 542 82
XM_017024790.1 XP_016880279.1 7 - 8 542 82
XM_017024791.1 XP_016880280.1 7 - 8 542 82
XM_017024792.1 XP_016880281.1 7 - 8 542 82
XM_017024793.1 XP_016880282.1 7 - 8 542 82
XM_017024794.1 XP_016880283.1 7 - 8 542 82
XM_017024795.1 XP_016880284.1 7 - 8 542 82
GenBank mRNA AF361219.1 - 8 - 9 530 82
AK025510.1 - 7 - 8 507 82
AK225757.1 - 7 - 8 550 82
AK314744.1 - 7 - 8 536 82
BC017390.1 - 7 - 8 547 82
BC117275.1 - 7 - 8 513 82
BC143386.1 - 7 - 8 513 82
BC143387.1 - 7 - 8 513 82
DA491709.1 - 7 - 8 528 82

View Full Product Details