Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
Chromosome Location
Chr.17: 60677811 - 61392838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099432.1 NP_001092902.1 15 - 16 1595 89
NM_001320470.1 NP_001307399.1 15 - 16 1595 89
NM_017679.3 NP_060149.3 15 - 16 1595 89
XM_005257472.1 XP_005257529.1 15 - 16 1546 89
XM_011524941.1 XP_011523243.1 15 - 16 1546 89
XM_017024786.1 XP_016880275.1 15 - 16 1546 89
XM_017024790.1 XP_016880279.1 15 - 16 1546 89
XM_017024791.1 XP_016880280.1 15 - 16 1546 89
XM_017024792.1 XP_016880281.1 15 - 16 1546 89
XM_017024793.1 XP_016880282.1 15 - 16 1546 89
XM_017024795.1 XP_016880284.1 15 - 16 1546 89
XM_017024796.1 XP_016880285.1 7 - 8 972 89
GenBank mRNA AF361219.1 - 16 - 17 1534 89
AK000135.1 - 1 - 2 73 89
AK022526.1 - 7 - 8 844 89
AK023054.1 - 7 - 8 942 89
AK025510.1 - 15 - 16 1511 89
AK225757.1 - 15 - 16 1554 89
AK314744.1 - 15 - 16 1540 89
AL831895.1 - 1 - 2 66 89
BC001250.1 - 1 - 2 82 89
BC117275.1 - 15 - 16 1517 89
BC143386.1 - 15 - 16 1517 89

View Full Product Details