Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 12132432-12132938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66D
Entrez Gene ID
100132923
Gene Name
family with sequence similarity 66 member D
Gene Aliases
-
UniGene
Hs.546711
Gene Chromosome Location
Chr.8: 12115782-12151189 on Build GRCh38
Transcripts
NR_027425.1 NM_001256869.1

Gene Symbol
USP17L7
Entrez Gene ID
392197
Gene Name
ubiquitin specific peptidase 17-like family member 7
Gene Aliases
-
UniGene
Hs.741132
Gene Chromosome Location
Chr.8: 12132417-12134009 on Build GRCh38
Transcripts
NR_027425.1 NM_001256869.1

SNP Detail

SNP ID Chromosome Location
rs552060040 Chr.8: 12132843-12132843 on Build GRCh38
rs767173107 Chr.8: 12132586-12132586 on Build GRCh38
rs763140876 Chr.8: 12132781-12132781 on Build GRCh38
rs751490883 Chr.8: 12132730-12132730 on Build GRCh38
rs751644125 Chr.8: 12132756-12132756 on Build GRCh38
rs777386482 Chr.8: 12132813-12132813 on Build GRCh38
rs528594703 Chr.8: 12132554-12132554 on Build GRCh38
rs747270552 Chr.8: 12132521-12132521 on Build GRCh38
rs773883622 Chr.8: 12132746-12132746 on Build GRCh38
rs373078712 Chr.8: 12132535-12132535 on Build GRCh38
rs575984940 Chr.8: 12132742-12132742 on Build GRCh38
rs554822461 Chr.8: 12132623-12132623 on Build GRCh38
rs752910672 Chr.8: 12132753-12132753 on Build GRCh38
rs757267356 Chr.8: 12132758-12132758 on Build GRCh38
rs777222260 Chr.8: 12132749-12132749 on Build GRCh38
rs781020687 Chr.8: 12132737-12132737 on Build GRCh38
rs762964876 Chr.8: 12132531-12132531 on Build GRCh38
rs751711218 Chr.8: 12132789-12132789 on Build GRCh38
rs754248696 Chr.8: 12132715-12132715 on Build GRCh38
rs759457495 Chr.8: 12132494-12132494 on Build GRCh38
rs769508744 Chr.8: 12132741-12132741 on Build GRCh38
rs138217195 Chr.8: 12132805-12132805 on Build GRCh38
rs763464728 Chr.8: 12132487-12132487 on Build GRCh38
rs758119917 Chr.8: 12132851-12132851 on Build GRCh38
rs530239874 Chr.8: 12132506-12132506 on Build GRCh38
rs756653576 Chr.8: 12132876-12132876 on Build GRCh38
rs756447152 Chr.8: 12132639-12132639 on Build GRCh38
rs769776265 Chr.8: 12132874-12132875 on Build GRCh38
rs775403371 Chr.8: 12132573-12132573 on Build GRCh38
rs755306820 Chr.8: 12132716-12132716 on Build GRCh38
rs780639913 Chr.8: 12132878-12132878 on Build GRCh38
rs779880641 Chr.8: 12132768-12132768 on Build GRCh38
rs534336835 Chr.8: 12132901-12132901 on Build GRCh38
rs774632310 Chr.8: 12132478-12132478 on Build GRCh38
rs754919341 Chr.8: 12132458-12132458 on Build GRCh38
rs774671797 Chr.8: 12132884-12132884 on Build GRCh38
rs765191501 Chr.8: 12132530-12132530 on Build GRCh38
rs763202606 Chr.8: 12132754-12132754 on Build GRCh38
rs745673298 Chr.8: 12132738-12132738 on Build GRCh38
rs754158929 Chr.8: 12132690-12132690 on Build GRCh38
rs770094674 Chr.8: 12132775-12132775 on Build GRCh38
rs761908940 Chr.8: 12132792-12132792 on Build GRCh38
rs143104523 Chr.8: 12132626-12132626 on Build GRCh38
rs753431189 Chr.8: 12132632-12132632 on Build GRCh38
rs200322250 Chr.8: 12132773-12132773 on Build GRCh38
rs748554467 Chr.8: 12132481-12132481 on Build GRCh38
rs772270937 Chr.8: 12132888-12132888 on Build GRCh38
rs752532647 Chr.8: 12132460-12132460 on Build GRCh38
rs777310148 Chr.8: 12132774-12132774 on Build GRCh38
rs761996642 Chr.8: 12132838-12132838 on Build GRCh38
rs752110423 Chr.8: 12132635-12132635 on Build GRCh38
rs542459665 Chr.8: 12132528-12132528 on Build GRCh38
rs759077967 Chr.8: 12132633-12132633 on Build GRCh38
rs9694759 Chr.8: 12132529-12132529 on Build GRCh38
rs181389292 Chr.8: 12132831-12132831 on Build GRCh38
rs748937319 Chr.8: 12132771-12132771 on Build GRCh38
rs747005763 Chr.8: 12132799-12132800 on Build GRCh38
rs777934376 Chr.8: 12132791-12132793 on Build GRCh38
rs773542601 Chr.8: 12132520-12132520 on Build GRCh38
rs181719434 Chr.8: 12132673-12132673 on Build GRCh38
rs777482158 Chr.8: 12132853-12132853 on Build GRCh38
rs528506213 Chr.8: 12132750-12132750 on Build GRCh38
rs774574096 Chr.8: 12132832-12132832 on Build GRCh38
rs141263690 Chr.8: 12132645-12132645 on Build GRCh38
rs765190409 Chr.8: 12132909-12132909 on Build GRCh38
rs771553045 Chr.8: 12132748-12132748 on Build GRCh38
rs747764168 Chr.8: 12132809-12132809 on Build GRCh38
rs779379530 Chr.8: 12132539-12132539 on Build GRCh38
rs779199706 Chr.8: 12132692-12132692 on Build GRCh38
rs765234783 Chr.8: 12132495-12132495 on Build GRCh38
rs72496213 Chr.8: 12132532-12132532 on Build GRCh38
rs750317099 Chr.8: 12132763-12132763 on Build GRCh38
rs547069010 Chr.8: 12132776-12132776 on Build GRCh38
rs765568954 Chr.8: 12132718-12132718 on Build GRCh38
rs760835837 Chr.8: 12132484-12132484 on Build GRCh38
rs779475319 Chr.8: 12132512-12132512 on Build GRCh38
rs769110625 Chr.8: 12132883-12132883 on Build GRCh38
rs758583219 Chr.8: 12132737-12132737 on Build GRCh38
rs749803310 Chr.8: 12132475-12132475 on Build GRCh38
rs530899859 Chr.8: 12132534-12132534 on Build GRCh38
rs749664876 Chr.8: 12132829-12132829 on Build GRCh38
rs768705010 Chr.8: 12132579-12132579 on Build GRCh38
rs750109653 Chr.8: 12132588-12132588 on Build GRCh38
rs778633367 Chr.8: 12132807-12132807 on Build GRCh38
rs768352653 Chr.8: 12132772-12132772 on Build GRCh38
rs148208217 Chr.8: 12132630-12132630 on Build GRCh38
rs772354326 Chr.8: 12132482-12132482 on Build GRCh38
rs746144415 Chr.8: 12132616-12132616 on Build GRCh38
rs191378034 Chr.8: 12132747-12132747 on Build GRCh38
rs773367053 Chr.8: 12132892-12132892 on Build GRCh38
rs777887550 Chr.8: 12132728-12132728 on Build GRCh38
rs766887803 Chr.8: 12132714-12132714 on Build GRCh38
rs760219926 Chr.8: 12132595-12132595 on Build GRCh38
rs764387803 Chr.8: 12132787-12132787 on Build GRCh38
rs781108720 Chr.8: 12132760-12132760 on Build GRCh38
rs765951433 Chr.8: 12132596-12132596 on Build GRCh38
rs754068779 Chr.8: 12132624-12132624 on Build GRCh38
rs778612463 Chr.8: 12132459-12132459 on Build GRCh38
rs771897307 Chr.8: 12132655-12132655 on Build GRCh38
rs563941093 Chr.8: 12132697-12132697 on Build GRCh38
rs752896850 Chr.8: 12132726-12132726 on Build GRCh38
rs536061475 Chr.8: 12132620-12132620 on Build GRCh38
rs770762436 Chr.8: 12132695-12132695 on Build GRCh38
rs765076098 Chr.8: 12132849-12132849 on Build GRCh38
rs761701570 Chr.8: 12132580-12132580 on Build GRCh38
rs776288222 Chr.8: 12132667-12132667 on Build GRCh38
rs770590166 Chr.8: 12132469-12132469 on Build GRCh38
rs758444853 Chr.8: 12132693-12132693 on Build GRCh38
rs762377861 Chr.8: 12132702-12132702 on Build GRCh38
rs748701722 Chr.8: 12132545-12132545 on Build GRCh38
rs760441634 Chr.8: 12132627-12132627 on Build GRCh38
rs748440093 Chr.8: 12132885-12132885 on Build GRCh38
rs570885648 Chr.8: 12132537-12132537 on Build GRCh38
rs746530084 Chr.8: 12132868-12132868 on Build GRCh38
rs746916783 Chr.8: 12132694-12132694 on Build GRCh38
rs763933510 Chr.8: 12132502-12132502 on Build GRCh38
rs990270586 Chr.8: 12132891-12132891 on Build GRCh38
rs748561283 Chr.8: 12132516-12132516 on Build GRCh38
rs747360618 Chr.8: 12132551-12132551 on Build GRCh38
rs757663451 Chr.8: 12132614-12132614 on Build GRCh38
rs749706852 Chr.8: 12132881-12132881 on Build GRCh38
rs752525914 Chr.8: 12132911-12132911 on Build GRCh38
rs753653976 Chr.8: 12132846-12132846 on Build GRCh38
rs78624849 Chr.8: 12132524-12132524 on Build GRCh38
rs776380717 Chr.8: 12132696-12132696 on Build GRCh38
rs758071712 Chr.8: 12132463-12132463 on Build GRCh38
rs761218472 Chr.8: 12132712-12132712 on Build GRCh38
rs752022254 Chr.8: 12132612-12132612 on Build GRCh38
rs769907288 Chr.8: 12132617-12132617 on Build GRCh38
rs368104450 Chr.8: 12132906-12132906 on Build GRCh38
rs762776638 Chr.8: 12132918-12132918 on Build GRCh38
rs780342645 Chr.8: 12132619-12132619 on Build GRCh38
rs763054272 Chr.8: 12132577-12132577 on Build GRCh38
rs766492898 Chr.8: 12132485-12132485 on Build GRCh38
rs529866508 Chr.8: 12132803-12132803 on Build GRCh38
rs756462032 Chr.8: 12132683-12132683 on Build GRCh38
rs13278959 Chr.8: 12132686-12132686 on Build GRCh38
rs755517325 Chr.8: 12132510-12132510 on Build GRCh38
rs576469109 Chr.8: 12132647-12132647 on Build GRCh38
rs763624920 Chr.8: 12132707-12132707 on Build GRCh38
rs1006694004 Chr.8: 12132654-12132654 on Build GRCh38
rs201590848 Chr.8: 12132691-12132691 on Build GRCh38
rs762444079 Chr.8: 12132672-12132672 on Build GRCh38
rs565827300 Chr.8: 12132603-12132603 on Build GRCh38
rs774978624 Chr.8: 12132670-12132670 on Build GRCh38
rs768103685 Chr.8: 12132744-12132744 on Build GRCh38
rs750879013 Chr.8: 12132678-12132678 on Build GRCh38
rs772445941 Chr.8: 12132519-12132519 on Build GRCh38
rs200816630 Chr.8: 12132825-12132825 on Build GRCh38
rs745374238 Chr.8: 12132509-12132509 on Build GRCh38
rs754673564 Chr.8: 12132806-12132806 on Build GRCh38
rs781390464 Chr.8: 12132615-12132615 on Build GRCh38
rs756018706 Chr.8: 12132804-12132804 on Build GRCh38
rs757181275 Chr.8: 12132735-12132735 on Build GRCh38
rs536984464 Chr.8: 12132637-12132637 on Build GRCh38
rs746442485 Chr.8: 12132814-12132814 on Build GRCh38
rs9694780 Chr.8: 12132799-12132799 on Build GRCh38
rs750744170 Chr.8: 12132638-12132638 on Build GRCh38
rs780637517 Chr.8: 12132507-12132507 on Build GRCh38
rs753341106 Chr.8: 12132597-12132597 on Build GRCh38
rs760814893 Chr.8: 12132894-12132894 on Build GRCh38
rs779854328 Chr.8: 12132877-12132878 on Build GRCh38
rs559508834 Chr.8: 12132498-12132498 on Build GRCh38
rs9694757 Chr.8: 12132477-12132477 on Build GRCh38
rs759369571 Chr.8: 12132905-12132905 on Build GRCh38
rs746793142 Chr.8: 12132657-12132657 on Build GRCh38
rs773450756 Chr.8: 12132483-12132483 on Build GRCh38
rs746622009 Chr.8: 12132467-12132467 on Build GRCh38
rs773280303 Chr.8: 12132839-12132839 on Build GRCh38
rs780829149 Chr.8: 12132474-12132474 on Build GRCh38
rs752890914 Chr.8: 12132600-12132600 on Build GRCh38
rs758438219 Chr.8: 12132727-12132727 on Build GRCh38
rs189370611 Chr.8: 12132668-12132668 on Build GRCh38
rs149455714 Chr.8: 12132559-12132559 on Build GRCh38
rs752447823 Chr.8: 12132850-12132850 on Build GRCh38
rs764145048 Chr.8: 12132755-12132755 on Build GRCh38
rs569861778 Chr.8: 12132828-12132828 on Build GRCh38
rs570065479 Chr.8: 12132848-12132848 on Build GRCh38
rs755920949 Chr.8: 12132765-12132765 on Build GRCh38
rs780432469 Chr.8: 12132640-12132640 on Build GRCh38
rs546566395 Chr.8: 12132743-12132743 on Build GRCh38
rs752814774 Chr.8: 12132854-12132854 on Build GRCh38
rs773610939 Chr.8: 12132711-12132711 on Build GRCh38
rs766331193 Chr.8: 12132900-12132900 on Build GRCh38
rs374095675 Chr.8: 12132456-12132456 on Build GRCh38
rs778176441 Chr.8: 12132547-12132547 on Build GRCh38
rs751309469 Chr.8: 12132504-12132504 on Build GRCh38
rs770692024 Chr.8: 12132664-12132664 on Build GRCh38
rs560546561 Chr.8: 12132812-12132812 on Build GRCh38
rs540682599 Chr.8: 12132656-12132656 on Build GRCh38

View Full Product Details