Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLSPN
Entrez Gene ID
63967
Gene Name
claspin
Gene Aliases
-
Chromosome Location
Chr.1: 35720465 - 35769985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190481.1 NP_001177410.1 11 - 12 2163 80
NM_022111.3 NP_071394.2 12 - 13 2355 80
XM_011541938.2 XP_011540240.1 12 - 13 2378 80
XM_011541939.2 XP_011540241.1 12 - 13 2372 80
XM_011541940.2 XP_011540242.1 12 - 13 2372 80
XM_011541941.2 XP_011540243.1 11 - 12 2210 80
GenBank mRNA AF297866.1 - 12 - 13 2355 80
BC113116.1 - 12 - 13 2348 80
BC115025.1 - 11 - 12 2156 80
BC115026.1 - 12 - 13 2348 80
BC137279.1 - 12 - 13 2334 80
BC140789.1 - 12 - 13 2334 80

View Full Product Details