Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HELB
Entrez Gene ID
92797
Gene Name
DNA helicase B
Gene Aliases
DHB, hDHB
Chromosome Location
Chr.12: 66302545 - 66343643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033647.4 NP_387467.2 1 - 2 245 150
NR_135080.1 1 - 2 245 150
NR_135081.1 1 - 2 245 150
XM_005269234.2 XP_005269291.1 1 - 2 223 150
XM_017020230.1 XP_016875719.1 1 - 2 223 150
GenBank mRNA AF319995.1 - 1 - 2 186 150
AK290894.1 - 1 - 2 210 150
BC143409.1 - 1 - 2 211 150
BC143411.1 - 1 - 2 211 150
DN997712.1 - 1 - 2 254 150

View Full Product Details