Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 61500384-61500894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
UniGene
Hs.332422
Gene Chromosome Location
Chr.8: 61500556-61714640 on Build GRCh38
Transcripts
NM_001164750.1 NM_004318.3 XM_005251235.2 XM_005251236.2 XM_005251238.2 XM_005251239.2 XM_005251240.1 XM_005251242.2 XM_005251243.2 XM_005251244.1 XM_005251246.2 XM_005251247.2 XM_005251248.1 XM_005251250.2 XM_017013419.1 XM_017013420.1 XM_017013421.1 XM_017013422.1 XM_017013423.1 XM_017013424.1 XM_017013425.1 XM_017013426.1 XM_017013427.1 XM_017013428.1 XM_017013429.1 XM_017013430.1 XM_017013431.1 XM_017013432.1 XM_017013433.1 XM_017013434.1 XM_017013435.1 XM_017013436.1 XM_017013437.1 XM_017013438.1 XM_017013439.1 XM_017013440.1 XM_017013441.1 XM_017013442.1 XM_017013443.1 XM_017013444.1 NM_173519.2 XM_017013141.1 XM_017013142.1

Gene Symbol
CLVS1
Entrez Gene ID
157807
Gene Name
clavesin 1
Gene Aliases
CRALBPL, RLBP1L1
UniGene
Hs.591874
Gene Chromosome Location
Chr.8: 61057158-61501645 on Build GRCh38
Transcripts
NM_001164750.1 NM_004318.3 XM_005251235.2 XM_005251236.2 XM_005251238.2 XM_005251239.2 XM_005251240.1 XM_005251242.2 XM_005251243.2 XM_005251244.1 XM_005251246.2 XM_005251247.2 XM_005251248.1 XM_005251250.2 XM_017013419.1 XM_017013420.1 XM_017013421.1 XM_017013422.1 XM_017013423.1 XM_017013424.1 XM_017013425.1 XM_017013426.1 XM_017013427.1 XM_017013428.1 XM_017013429.1 XM_017013430.1 XM_017013431.1 XM_017013432.1 XM_017013433.1 XM_017013434.1 XM_017013435.1 XM_017013436.1 XM_017013437.1 XM_017013438.1 XM_017013439.1 XM_017013440.1 XM_017013441.1 XM_017013442.1 XM_017013443.1 XM_017013444.1 NM_173519.2 XM_017013141.1 XM_017013142.1

SNP Detail

SNP ID Chromosome Location
rs1011175287 Chr.8: 61500429-61500429 on Build GRCh38
rs533453926 Chr.8: 61500541-61500541 on Build GRCh38
rs572448086 Chr.8: 61500776-61500777 on Build GRCh38
rs879648785 Chr.8: 61500629-61500630 on Build GRCh38
rs924393699 Chr.8: 61500480-61500480 on Build GRCh38
rs962394840 Chr.8: 61500415-61500415 on Build GRCh38
rs986220705 Chr.8: 61500511-61500511 on Build GRCh38
rs73685070 Chr.8: 61500748-61500748 on Build GRCh38
rs59691966 Chr.8: 61500643-61500646 on Build GRCh38
rs540541641 Chr.8: 61500474-61500477 on Build GRCh38
rs550151793 Chr.8: 61500663-61500663 on Build GRCh38
rs977129567 Chr.8: 61500454-61500454 on Build GRCh38
rs1017662324 Chr.8: 61500686-61500686 on Build GRCh38
rs200668110 Chr.8: 61500643-61500643 on Build GRCh38
rs1043782335 Chr.8: 61500617-61500617 on Build GRCh38
rs190359574 Chr.8: 61500619-61500619 on Build GRCh38
rs1056678065 Chr.8: 61500652-61500652 on Build GRCh38
rs576789401 Chr.8: 61500685-61500686 on Build GRCh38
rs77497713 Chr.8: 61500440-61500441 on Build GRCh38
rs912079459 Chr.8: 61500527-61500527 on Build GRCh38
rs902679874 Chr.8: 61500659-61500660 on Build GRCh38
rs1047802219 Chr.8: 61500557-61500557 on Build GRCh38
rs748499262 Chr.8: 61500456-61500456 on Build GRCh38
rs184558557 Chr.8: 61500569-61500569 on Build GRCh38
rs553943290 Chr.8: 61500645-61500646 on Build GRCh38
rs987160993 Chr.8: 61500498-61500498 on Build GRCh38
rs564281291 Chr.8: 61500605-61500605 on Build GRCh38
rs770185483 Chr.8: 61500641-61500642 on Build GRCh38
rs1019809740 Chr.8: 61500754-61500754 on Build GRCh38
rs988396036 Chr.8: 61500583-61500583 on Build GRCh38
rs771491905 Chr.8: 61500656-61500656 on Build GRCh38
rs555812226 Chr.8: 61500452-61500452 on Build GRCh38
rs929146206 Chr.8: 61500551-61500551 on Build GRCh38
rs768273220 Chr.8: 61500445-61500446 on Build GRCh38
rs35458405 Chr.8: 61500589-61500590 on Build GRCh38
rs563014370 Chr.8: 61500504-61500504 on Build GRCh38
rs946695744 Chr.8: 61500615-61500615 on Build GRCh38
rs928035231 Chr.8: 61500798-61500798 on Build GRCh38
rs1055559787 Chr.8: 61500622-61500622 on Build GRCh38
rs34833311 Chr.8: 61500723-61500724 on Build GRCh38
rs868376881 Chr.8: 61500484-61500484 on Build GRCh38
rs914150846 Chr.8: 61500833-61500833 on Build GRCh38
rs148273510 Chr.8: 61500642-61500645 on Build GRCh38
rs973734273 Chr.8: 61500472-61500472 on Build GRCh38
rs540892468 Chr.8: 61500419-61500419 on Build GRCh38
rs546525365 Chr.8: 61500653-61500653 on Build GRCh38
rs567328032 Chr.8: 61500825-61500826 on Build GRCh38
rs943258887 Chr.8: 61500535-61500536 on Build GRCh38
rs914161787 Chr.8: 61500590-61500590 on Build GRCh38
rs181087874 Chr.8: 61500829-61500829 on Build GRCh38
rs765253006 Chr.8: 61500770-61500775 on Build GRCh38
rs767643456 Chr.8: 61500864-61500865 on Build GRCh38
rs769699906 Chr.8: 61500648-61500649 on Build GRCh38
rs966218782 Chr.8: 61500796-61500796 on Build GRCh38
rs4507756 Chr.8: 61500661-61500661 on Build GRCh38
rs748828747 Chr.8: 61500727-61500727 on Build GRCh38
rs939408980 Chr.8: 61500837-61500837 on Build GRCh38
rs148048456 Chr.8: 61500845-61500846 on Build GRCh38
rs964418763 Chr.8: 61500705-61500705 on Build GRCh38
rs895161454 Chr.8: 61500648-61500648 on Build GRCh38
rs926659148 Chr.8: 61500639-61500639 on Build GRCh38
rs988753674 Chr.8: 61500824-61500824 on Build GRCh38
rs561627923 Chr.8: 61500660-61500660 on Build GRCh38
rs777955398 Chr.8: 61500622-61500623 on Build GRCh38
rs939341891 Chr.8: 61500578-61500578 on Build GRCh38
rs73685069 Chr.8: 61500683-61500683 on Build GRCh38
rs201416786 Chr.8: 61500645-61500645 on Build GRCh38
rs778153787 Chr.8: 61500491-61500491 on Build GRCh38
rs757129456 Chr.8: 61500593-61500593 on Build GRCh38
rs968297214 Chr.8: 61500846-61500846 on Build GRCh38
rs752800147 Chr.8: 61500573-61500574 on Build GRCh38
rs920391378 Chr.8: 61500547-61500547 on Build GRCh38
rs747498217 Chr.8: 61500595-61500595 on Build GRCh38
rs973691100 Chr.8: 61500728-61500728 on Build GRCh38
rs377236781 Chr.8: 61500440-61500440 on Build GRCh38
rs144738254 Chr.8: 61500486-61500486 on Build GRCh38
rs779198308 Chr.8: 61500571-61500571 on Build GRCh38
rs1027945604 Chr.8: 61500664-61500664 on Build GRCh38
rs759590717 Chr.8: 61500852-61500853 on Build GRCh38
rs554047609 Chr.8: 61500499-61500499 on Build GRCh38
rs200653464 Chr.8: 61500435-61500436 on Build GRCh38

View Full Product Details