The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144663.1 | NP_653264.1 | 1 - 2 | 140 | 146 |
GenBank mRNA | AF439154.1 | - | 1 - 2 | 140 | 146 |