Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMC4
Entrez Gene ID
147798
Gene Name
transmembrane channel like 4
Gene Aliases
-
Chromosome Location
Chr.19: 54160095 - 54173250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145303.2 NP_001138775.2 2 - 3 445 86
NM_144686.3 NP_653287.2 2 - 3 427 86
XM_011526486.2 XP_011524788.1 2 - 3 410 86
GenBank mRNA AK314973.1 - 2 - 3 348 86
AY236492.1 - 2 - 3 444 86
AY263166.1 - 2 - 3 324 86
BC025323.1 - 2 - 3 334 86
BQ082417.1 - 2 - 3 337 86

View Full Product Details